• USA Home
  • Search Results

  • 44dihydroxydiphenylsulfonebisfluorosulfinate414384215898511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4,4′-Dihydroxydiphenyl sulfone bisfluorosulfinate

1 Products
Image of 4,4′-Dihydroxydiphenyl sulfone bisfluorosulfinate
CAS Number:
Empirical Formula (Hill Notation):
 C12H8F2O8S3
Molecular Weight:
 414.38
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More