• USA Home
  • Search Results

  • bacitracin142269140587411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bacitracin

4 Products
Image of Bacitracin
CAS Number:
Empirical Formula (Hill Notation):
 C66H103N17O16S
Molecular Weight:
 1422.69
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Mode of Action
  Activity Spectrum
  Physical Form
  Special Grade
  Solubility
Show More