• USA Home
  • Search Results

  • doxorubicinhydrochloride579982531640911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Doxorubicin hydrochloride

5 Products
Synonym:   DOX, Hydroxydaunorubicin hydrochloride
Image of Doxorubicin hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C27H29NO11 · HCl
Molecular Weight:
 579.98
 Beilstein Registry Number:
 4229251
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Anal. Standard Form
  Activity Spectrum
  Assay
  Mode of Action
  Special Grade
  Physical Form
Show More