• USA Home
  • Search Results

  • erythromycin7339311407811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Erythromycin

8 Products
Image of Erythromycin
CAS Number:
Empirical Formula (Hill Notation):
 C37H67NO13
Molecular Weight:
 733.93
 Beilstein Registry Number:
 75279
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Suitability
  Potency
  Melting Point
  Anal. Standard Form
  Special Grade
  Color
  Mode of Action
  Activity Spectrum
  Physical Form
Show More