• USA Home
  • Search Results

  • ironoxideiiiiimagneticnanoparticlessolution23153131761911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iron oxide(II,III), magnetic nanoparticles solution

15 Products
Synonym:   Magnetic iron oxide nanocrystals, Magnetite, Superparamagnetic iron oxide nanoparticles
Image of Iron oxide(II,III), magnetic nanoparticles solution
CAS Number:
Empirical Formula (Hill Notation):
 Fe3O4
Molecular Weight:
 231.53
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Particle Dimension
  Functional group
  Color
  Physical Form
  Miscellaneous Specifications
  Concentration
  Density
Show More

Product #
Description
700312 Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene,
700304 Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene,
700320 Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene,
725358 Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O,
725331 Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O,
725366 Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O,
747327 Iron oxide(II,III), magnetic nanoparticles solution, 30 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion,
747424 Iron oxide(II,III), magnetic nanoparticles solution, 10 nm diameter, biotin functionalized, 1 mg/mL Fe, dispersion in H2O,
747254 Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL Fe in H2O, dispersion,
747343 Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion,
747319 Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion,
747300 Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion,
747408 Iron oxide(II,III), magnetic nanoparticles solution, 30 nm avg. part. size (TEM), PEG functionalized, dispersion, 1 mg/mL Fe in H2O,
747416 Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), biotin functionalized, 1 mg/mL Fe in H2O, dispersion,
790508 Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion,