• USA Home
  • Search Results

  • mitomycincfromstreptomycescaespitosus334335007711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mitomycin C from Streptomyces caespitosus

4 Products
Image of Mitomycin C from <I>Streptomyces caespitosus</I>
CAS Number:
Empirical Formula (Hill Notation):
 C15H18N4O5
Molecular Weight:
 334.33
 Beilstein Registry Number:
 7231816
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Physical Form
  Biological Source
  pK-Values
  Suitability
  Assay
  Color
  Solubility
  Mode of Action
Show More