• USA Home
  • Search Results

  • norcamphor1101549738111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Norcamphor

1 Products
Synonym:   2-Norbornanone, Bicyclo[2.2.1]heptan-2-one
Image of Norcamphor
CAS Number:
Empirical Formula (Hill Notation):
 C7H10O
Molecular Weight:
 110.15
 Beilstein Registry Number:
 1209657
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
N32601 Norcamphor, 98%,