• USA Home
  • Search Results

  • sorbitansesquioleate12345800743011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Sorbitan sesquioleate

1 Products
Synonym:   Arlacel® 83, Span® 83
Image of Sorbitan sesquioleate
CAS Number:
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More