• USA Home
  • Search Results

  • trichloroaceticacid163397603911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trichloroacetic acid

8 Products
Synonym:   TCA
Image of Trichloroacetic acid
CAS Number:
Linear Formula:
 Cl3CCOOH
Molecular Weight:
 163.39
 Beilstein Registry Number:
 970119
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  UV-Absorption Values
  Titertest
  Restriction
  Quality clarifier
  Purity Grade
  Product Line
  Special Grade
  Solubility
Show More