• USA Home
  • Search Results

  • tritonx10012345900293111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triton X-100

10 Products
Synonym:   t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether, Polyoxyethylene (10) isooctylphenyl ether
Image of Triton<SUP>™</SUP> X-100
CAS Number:
Linear Formula:
 4-(C8H17)C6H4(OCH2CH2)nOH, n~10
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Manufacturer designation
  Ice Shipping
  Absorption
  Product Line
  Package quantity
  Density
  Special Grade
  Solubility
  ID clarifier
Show More