• USA Home
  • Search Results

  • waterdeuteriumdepleted1802773218511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Water, deuterium-depleted

1 Products
Synonym:   Deuterated depleted water
Image of Water, deuterium-depleted
CAS Number:
Linear Formula:
 H2O
Molecular Weight:
 18.02
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
Show More