Chemistry > Aldrich - Chemistry > Technical Bulletins > AL 143 > Activated Carbon
Aldrich
Activated Carbon
TechBulletin3 AL-143
Mineral Adsorbents, Filter Agents and Drying Agents
Home / Molecular Sieves / Activated Alumina / Activated Carbon / Diatomaceous Earth
Ascarite II® Adsorbents / Montmorillonites and other Mineral Adsorbents / Drying Agents
Toxicity and Handling / References
III. ACTIVATED CARBON
Activated carbon is available in a variety of grades for purification and, particularly, for decoloration applications, as well as for specialized uses.
Darco® KB and KB-B activated carbon
These grades have exceptionally high adsorptive capacities for specific color bodies, making them particularly useful for purifying highly colored liquids. Darco KB-B is processed to achieve an exceptionally low iron content (100 ppm maximum extractable) for very sensitive applications.
Table 6 - Specifications for Activated Carbon
Catalog Number 16,155-1 24,223-3 29,259-1 24,224-1 24,226-8 24,227-6 26,001-0 27,809-2 27,810-6
Type Darco® Darco Darco Darco
G-60
Norit A® Darco
KB
Darco
KB-B
Description acid-washed lignite carbon, granular granular acidwashed, granular unwashed lignite carbon, granular high-purity powder powder wet powder wet powder
Particle size (mesh) 4-12 4-14 12-20 12-40 100-325 200-325 100-325 100-325
Surface area
(m2/g, approx.)
600 600 600 750 1500 1500
Pore volume (ml/g, dry basis) 0.95 0.95 0.95 0.95 2 2
Moisture
(%, max.)
12 2 33 33
Iodine number 800
Methylene Blue adsorption
(g/100g, min.)
15 12.5
pH (water extract, approx.) 6-8 9 5 5
Ash (%) 3.5 6 3 3
Molasses decolorizing number 440 185 185
Water solubles
(%, max.)
0.5 1.5 1.5
Tamped bulk density
(lb/cu ft)
25 a 28 28
Fe
(ppm, max.)
200 100
Phosphates (%, extractable) 0.5 0.5
Application decolorizing general general- purpose general- purpose high-purity use decolorizing b b
apacked, 0.460g/ml b described above
®Registered trademarks of American Norit Co., Inc.