Aldrich
Ascarite II Adsorbents
|
V. ASCARITE II®ADSORBENTS
|
|
Ascarite II is the second generation of a traditional brand of carbon dioxide adsorbents. Whereas the original Ascarite was derived from granular asbestos, Ascarite II is a sodium hydroxidecoated silica. It rapidly and quantitatively adsorbs CO2, (and acid gases), and is useful in a number of analytical and microanalytical procedures, physiological studies, etc.
The materials are self-indicating, gradually changing color (to white) within a narrow zone due to the formation of sodium carbonate.
|
| Table 8 - Ascarite II. Adsorbents |
| Catalog No. |
Grade |
Type |
Approx. particle size (mesh) |
| 22,391-3 |
Ascarite II |
granular |
820 |
| 22,392-1 |
Ascarite II |
granular |
2030 |
|
®Registered trademark of Arthur H. Thomas Co.