Sigma-Aldrich
Chemistry > Aldrich - Chemistry > Technical Bulletins > AL 143 > Diatomaceous Earth
Aldrich
Diatomaceous Earth
TechBulletin3 AL-143
Mineral Adsorbents, Filter Agents and Drying Agents
Home / Molecular Sieves / Activated Alumina / Activated Carbon / Diatomaceous Earth
Ascarite II® Adsorbents / Montmorillonites and other Mineral Adsorbents / Drying Agents
Toxicity and Handling / References
IV. DIATOMACEOUS EARTH
Known by a variety of tradenames, diatomaceous-earth products find common use as filter agents and all-purpose adsorbents, as well as in specialized applications, such as catalyst carriers. These products, also known as diatomaceous silicas, are powders generally having a silicon dioxide assay of about 90%.
The CAFA (Celite® Analytical Filter Aid) product is double-acid-washed to remove trace metal oxides and contains about 97.5% SiO2. It is useful as an analytical filter agent and for high-purity applications.
Celite 521 is acid-washed and contains about 94% SiO2. It is suitable for use where exceptional purity is desired.
The Celatom® products are flux-calcined, with a pH (10% slurry) of 8.0-9.5, and specific gravity of 2.33.
Table 7
Catalog No. 16,743-6 22,179-1 24,331-0 24,332-9 24,333-7 24,334-5
Grade CAFA Celite Celatom
FW-14
Celatom
FW-50
Celatom
FW-60
Celatom
FW-80
Flow rate 100 200 700 2500 3000 5500
Median size (µ) 2.5 3.5 — — — —
Clarity 1000 500 969 940 936 927
Acid-soluble iron (ppm) 60 15 — — — —
Wet density
(lb/cu ft)
17 19-90 19 18 18 17
Avg. particle size (µ) 7.5 14 8 15 19 23
Moisture (%) 1 0.5 — — — —
Color white-grey/ pink light-pink
to tan
white white white white
Celite is a registered trademark of JohnsManville Corp.
Celatom is a registered trademark of EaglePicher Industries, Inc.