Aldrich
Handling
Technical Bulletin banner
AL-209 (Europe only)
UN-Pac Packing System and Transfer Procedure for Type DS
Introduction / Specifications / Handling / Recommended Transfer Procedure
Storage / Container Return / Equipment
Handling:
The user must be a fully qualified and experienced laboratory or chemical production worker to handle these reagents. All users must be aware of the hazardous nature of many of these products. The material safety data sheet (MSDS), which is provided with each order, must be read and understood by the user prior to starting any work with the product contained in a UN-Pac container. In general, handle these products only under an inert atmosphere and exercise caution to prevent inhalation of vapors or direct contact with the skin. Operators must wear appropriate clothing with long sleeves, chemical resistant gloves, goggles, and full faceshield. Additional personal protective equipment must be used as required. In case of contact, immediately flush eyes or skin with water for at least 15 minutes while removing contaminated clothing and shoes. Seek medical attention promptly.
In case of fire, use a dry chemical extinguisher; do not use water with moisture reactive products. Store UN-Pac containers upright in a cool location, away from heat and direct sunlight. Refrigerate if necessary.
During all nitrogen-pressure transfer, the flowing liquid can generate a static charge. Therefore, the UN-Pac container and receiver must be connected to a suitable ground. Aldrich offers a 6-ft length of flat braided bare cable with copper clips at each end which makes a convenient grounding strap (Z15,030-4).
Technical experts are available by telephone to answer your questions regarding the proper handling of products packaged in UN-Pac containers. Contact Aldrich immediately if an inadvertent deviation from the recommended transfer procedure occurs during use of a UN-Pac container.
Alogo1 chemists helping chemists in research and industry
P.O. Box 355, Milwaukee, WI 53201 USA Telephone: 414-273-3850 • 800-558-9160
Fax: 414-273-4979 • 800-962-9591 TWX: 910-262-3052 Telex: 26 843
This information is presented to assist you in evaluating our product. It is intended for use by technically-skiIled persons. We do not guarantee favorable results, and we assume no liability inconnection with its use. This information is not intended as a license to operate under, or arecommendation to infringe, any patent covering any material or use. Not for drug, food, cosmetic or household use.