Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Fluorous Chemistry > Proteomics & Metabolomics
Fluorous Chemistry
Proteomics & Metabolomics
The seminal publication in this field was published by Peters et al from the Genomics Institute of the Novartis Research Foundation in 2005 and describes the use of fluorous bioreagents in proteomics. Within that publication, they described fluorous reagents for Cys tagging, fluorous thiols for phosphoproteome analysis, and fluorous NHS esters for amine tagging. Since that publication, the line of bioreagents has been extended to include products for carbonyl tagging and click chemistry.
Recommended Literature
Brittain, S. M.; Ficarro, S. B.; Brock, A.; Peters, E. C. Enrichment and Analysis of Peptide Subsets using Fluorous Affinity Tags and Mass Spectrometry. Nat. Biotechnol. 2005, 23, 463.
Fluorous maleimide
FTI Products: BR013173 (19425), BR107173 (40889)
Fluorous maleimide Structure Image
Fluorous maleimide is used for the tagging and enrichment of cysteinyl peptides.
Fluorous iodoacetamide
FTI Products: BR013174 (51526), BR107174 (55266)
Fluorous iodoacetamide Structure Image
Fluorous iodoacetamide is used for the tagging and enrichment of cysteinyl peptides.
Fluorous NHS ester
FTI Products: BR013175 (41687), BR107175 (73028)
Fluorous NHS ester Image
Fluorous NHS ester is used for the tagging of free amines such as peptide N-termini and lysines.
Fluorous azide
FTI Products: BR017190 (97087)
Fluorous NHS ester Image
Fluorous azide is used to conduct click chemistry in the tagging of alkyne modified biomolecules.
back to top