Sigma-Aldrich
Chemistry > Chemical Synthesis > Learning Center > Aldrich ChemFiles > ChemFiles 2001-2003 > Vol 2 No 1
ChemFilesTitle Vol. 2, No. 1
Products for Suzuki Coupling
Download PDF (1.03 MB)
Introduction / Boronic Acids / Boronic Acids Esters / Diboron Esters / Prolines & Pyroglutamates
Transition-Metal Catalysts / Palladium Scavengers / Large-Scale Suzuki Coupling / Reference Books
Suzuki-CoverEssential Tools for Today's Synthetic Chemist
The synthesis of biaryl compounds by reaction of aryl halides with arylboronic acids, commonly referred to as the Suzuki coupling,1 is an important area of growth. Of the wide variety of cross-coupling reactions, Suzuki coupling is the most general and widely used. Arylboronic acids are the favored reagents due to their stability, low toxicity, and limited side reactions. This brochure contains a comprehensive selection of boronic acids, boronic acid esters, diboron esters, and transition-metal catalysts useful for the Suzuki coupling reaction. New offerings are added monthly; if you do not see the material you need for your research, please call us at 1-800-231-8327 (USA) or your local office. Your new product suggestions, as always, are welcome and appreciated.