|
| |
Vol. 2, No. 1 Products for Suzuki Coupling
Download PDF (1.03 MB) Introduction / Boronic Acids / Boronic Acids Esters / Diboron Esters / Prolines & Pyroglutamates Transition-Metal Catalysts / Palladium Scavengers / Large-Scale Suzuki Coupling / Reference Books Palladium Scavengers
Scavenger resins have been commercially available for several years. Most of these scavengers have been used to sequester organic functionalities from solution in order to ease the purification process upon completion of the synthesis. Now, Aldrich is introducing a new class of quenching reagents: palladium scavengers on silica gel.
 |
53,792-6 3-(Diethylenetriamino)propyl-functionalized silica gel 5g; 25g; 100g |
 |
53,808-6 3-Mercaptopropyl-functionalized silica gel 5g; 25g; 100g |
Conditions: In separate flasks, two solutions of Pd(OAc) 2 (1000 ppm Pd) were prepared in THF. While stirring at room temperature, four equivalents of each of the silica-based scavengers were added to their respective flasks. As illustrated in the graph, the palladium was successfully scavenged from the solution within minutes. Simple filtration of the bound palladium allows for its recovery.
For questions regarding these scavengers or other resins, please contact the product manager at 414-298-6330, or via e-mail at bwandler@sial.com
If you would like more information regarding supported reagents, request your 2001-2002 Sigma-Aldrich Combinatorial Chemistry catalog today. This catalog features everything you need for polymer-assisted organic synthesis, parallel synthesis, and high-throughput screening.
Larger quantities are available from SiliCycle, Inc.