Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
ChemFilesTitle Vol . 2, No. 5
Rieke® Organozinc & Organomagnesium Reagents
Download PDF (938 KB)
Introduction / Rieke® Organozinc Reagents / Rieke® Organomagnesium Reagents
Rieke® Highly Reactive Metals / Rieke® Specialty Conducting Polymers
Aldrich Grignards / Lab Equipment / Reference Books
Rieke® Highly Reactive Metals
Metal-enhanced organic synthesis via organometallic intermediates is a widely used preparative route for thousands of organic compounds. However, in some cases, ordinary bulk metal forms fail to react with organic substrates. Rieke® Highly Reactive Magnesium (Mg*) and Rieke® Highly Reactive Zinc (Zn*) react efficiently with a variety of organic substrates.
Cat. No. Product Name Unit Size
49,957-9 Magnesium, highly reactive Rieke® metal
(2.5g Mg* in 100mL tetrahydrofuran)
100mL
49,955-2 Zinc, highly reactive Rieke® metal
(5g Zn* in 100mL tetrahydrofuran)
100mL