Sigma-Aldrich
Chemistry > Chemical Synthesis > Learning Center > Aldrich ChemFiles > ChemFiles 2001-2003 > Vol 2 No 6
ChemFilesTitle Vol. 2, No. 6
Functionalized SILICA GELS for Organic Synthesis
Download PDF (805 KB)
Introduction / Quality Control / Organic Synthesis / Scavengers
Supported Reagents / Linker Resins / Kits / References
SilicaGelsCover
Introduction
Sigma-Aldrich and SiliCycle Inc. have partnered to offer you a wide-range of functionalized silica gels for organic synthesis. SiliCycle Inc. manufacture these novel products to combine with the 700+ resins that Sigma-Aldrich now offers to help your synthetic needs.
These novel silica gel products offer numerous advantages over the well-known gel-type resins.
Advantages of the Inert Support
  • Solvent independent
    • Rigid porous structure
      • No swelling
      • Fast kinetics
      • Can be used in large-scale purification (discovery to process)
    • No low molecular weight residues
      • High purity
  • No non-specific binding
    • High Yield
  • High density of functional groups
    • Small volume of silica gel required
This ChemFile details the complete range of functionalized silica gels and their applications. All of these products are available in pack sizes of 5 grams to multi-kilos.