|
| |
Vol. 2, No. 8 New Nitrogen-Containing Heterocycles
Introduction / 5-Membered Heterocyclic Rings / 6-Membered Heterocyclic Rings Mitsunaobu Reagents / Polymer-Supported Reagents / JandaJels™ / Functionalized Silica Gels JandaJels™
These novel insoluble supports contain a flexible tetrahydrofuran-derived cross-linker that offers increased swelling/solvation in common solvents used in solid-phase organic synthesis and increased homogeneity over other commercially available polystyrene resins.
57,638-7
JANDAJEL™ POLYPYRIDINE, 100-200 MESH (PYRIDINE ON JANDAJEL™)
Supported catalyst.
5g; 25g
|
57,829-0
JANDAJEL™ POLYPYRIDINE, 50-100 MESH (PYRIDINE ON JANDAJEL™)
Supported catalyst.
5g; 25g
|
57,049-4
JANDAJEL™ - 1,3,4,6,7,8-HEXAHYDRO-2H-PYRIMIDO-[1,2-A]PYRIMIDINE (TBD ON JANDAJEL™)
Supported scavenger for phenols and peroxides.
5g; 25g