Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
ChemFilesTitle Vol. 2, No. 8
New Nitrogen-Containing Heterocycles
Introduction / 5-Membered Heterocyclic Rings / 6-Membered Heterocyclic Rings
Mitsunaobu Reagents / Polymer-Supported Reagents / JandaJels™ / Functionalized Silica Gels
Mitsunaobu Reagents
One of the most powerful and widely used carbon-carbon bond forming reactions in organic synthesis is the Mitsunobu reaction. The Mitsunobu reaction also can be applied to the preparation of other moieties, such as N-alkylamides or imides.
56,311-0  NEW
DIETHYL AZODICARBOXYLATE, 40 wt.% solution in toluene, (DEAD solution)
563110The most common reagent used in Mitsunobu reactions, now available once again as a stable toluene solution.
50g; 250g
Other Azodicarboxylates used for the Mitsunobu reaction:
13599213,599-2
DI-TERT-BUTYL AZODICARBOXYLATE, 98%, (DBAD)
5g
22554122,554-1
DIISOPROPYL AZODICARBOXYLATE, 95% (DIAD)
5g; 100g
25592025,592-0
1,1'-(AZODICARBONYL)DIPIPERIDINE, 99%
1g; 5g; 25g