Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Dimeric Phosphazine Bases
Vol. 3, No. 1
Strong and Hindered Bases in Organic Synthesis
Download a PDF version
Introduction / Phosphazene Bases / Verkade's Superbases / Traditional bases
Preservation of Reagents / Aldrich Schlenk-Type Glassware
 Phosphazene Bases Introduction / Monomeric Phosphazene Bases (P1)
Dimeric Phosphazene Bases (P2) / Tetrameric Phosphazene Bases (P4)
Dimeric Phosphazene Bases (P2)
Aldrich lists two P2 phosphazene bases (79416 and 79417), which differ with respect to their steric hindrance and basicity (Table 3).
Table 3. Order of Basicity of P2 Bases
Order of steric hindrance: P2-t-Bu >> P2-t-Et
Aldrich Product Number: 79416   79417
Basicity (MeCNpKBH+): 33.5   32.9
The phosphazene base P2-t-Bu is a solid at room temperature (m.p. 76 0C). and is available as a 2 M solution in THF (79416). P2-Et (79417) is offered as a stable, distillable liquid, which can easily be handled with syringe techniques. Both exhibit characteristics similar to those of DBU, but compared to DBU they are more hindered, much less sensitive towards hydrolysis, and 8.5-9 orders of magnitude more basic[2,3] See Table 4 for a list of our P2 bases with their properties, package size etc.
Table 4. Phospazene bases P2
79416
Phosphazene base P2-t-Bu solution
1-tert-Butyl-2,2,4,4,4-pentakis(dimethylamino)-2L5,4L5-catenadi(phosphazene)
tert-Butylimino-tris(dimethylamino)phosphorane
purum, ~2.0 M in THF   C14H39N7P2   Mr 367.5   [111324-03-9]   1 mL, 5 mL, 25 mL
79417
Phosphazene base P2-Et
1-Ethyl-2,2,4,4,4-pentakis(dimethylamino)-2L5,4L5-catenadi(phosphazene)
purum, 98.0% (NT)   C12H35N7P2   Mr 339.4   [165535-45-5]   1 mL, 5 mL
Applications: P2-Et 79417 is particularly suitable for E2-eliminations[3,4,5,6] being around 4 orders of magnitude more reactive than DBU. Deprotonations for spectroscopic investigations,[1] double bond shifts in vinyl sulfones,[7] and alkyl-lithium activation[8] have also been reported. The P2-t-Bu (79416) is particularly suitable for deprotonation (activation) of pronucleophiles. The P2 bases have attracted interest in the field of anionic polymerisation[8,9] and have been reported to support alkylation-,[10,11,12,13] michael addition-,[13,14] aldol condensation-,[13,15,16] and copper-induced coupling reactions.[17] Among the successfully converted low acidic substrates are alcohols,[10] phenols,[5] carbamates,[11] NH-acidic heterocycles,[6] phosphines,[12] benzyl sulfones,[13] glycin ester Schiff-bases,[14,15] a-azidoketones,[18] and N-alkylpyridinium salts.[3] Cinnamic esters,[13] chiral phenyloxiranes,[16] chiral amino acids,[14,15] biaryl thioethers,[17] and pyrazines[18] are thus efficiently synthesized.
References:
  1. Schwesinger, R. et al., Liebigs Ann. 1996, 1055.
  2. Angelaud, R., Landais, Y., Tetrahedron Lett. 1997, 38, 8841.
  3. Dega-Szafran, Z. et al., J. Phys. Org. Chem. 1999, 12, 39.
  4. Prinzbach, H. et al., Tetrahedron Lett. 1987, 28, 2517.
  5. Zundel, G. et al., J. Phys. Chem. 1996, 100, 9021; Sobczyk, L. et al., J. Chem. Res. (S) 1997, 151.
  6. Zundel, G. et al., J. Mol. Struct. 1997, 436, 103; Bauer, R., Zundel, G., J. Phys. Chem. A 2002, 106, 5828.
  7. Fuchs, P. L. et al., Tetrahedron Lett. 1996, 37, 5249; Fuchs, P. L. et al., Tetrahedron Lett. 1997, 38, 2601.
  8. Mvller, M. et al., Macromol. Symp. 1996 (Intern. Symposium on Ionic Polymerization 1995), 331; Molenberg, A., Mvller, M., Macromol. Chem. Phys. 1997, 198, 717.
  9. Memeger Jr., W. et al., Macromolecules 1996, 29, 6475, 8568.
  10. Schwesinger, R., Nachr. Chem. Tech. Lab. 1990, 38, 1214.
  11. Prinzbach, H. et al., Angew. Chem. Int Ed. Engl. 1989, 28, 1253.
  12. Uhlig, F. et al., Phosphorus, Sulfur & Silicon 1993, 81, 155; Uhlig, F. et al., Phosphorus, Sulfur & Silicon 1993, 84, 181.
  13. Najera, C. et al., J. Org. Chem. 2002, 67, 5216.
  14. Solladii-Cavallo, A. et al., Synlett. 1997, 217.
  15. Solladii-Cavallo, A. et al., Synlett. 2000, 327.
  16. Solladii-Cavallo, A. et al., Eur. J. Org. Chem. 2000, 1077; Solladii-Cavallo, A. et al., Tetrahedron Lett. 2000, 41, 7309.
  17. Palomo, C. et al., Tetrahedron Lett. 2000, 41, 1283.
  18. Winterfeldt, E. et al., Eur. J. Org. Chem. 1998, 2811.