|
Introduction / Phosphazene Bases / Verkade's Superbases / Traditional bases Preservation of Reagents / Aldrich Schlenk-Type Glassware / Miscellaneous Information
Verkade's bases
Verkade's bases, football-shaped proazaphosphatrane molecules of type 1, are very strong bases due to the extraordinary stability of 2 when 1 reacts with a proton.
Due to the stability of the protonated form 2, Verkade's bases are about eight orders of magnitude stronger as a Lewis base than any amine known, including the prominently used DBU, DBN and Proton Sponge. (14795).
Applications: Verkade's bases have been successfully applied in a variety of organic reactions, such as alkylations, dehydrohalogenations, acylations,[1] a variety of condensation and organometallic reactions for carbon-carbon bond formation.[2] A second characteristic of the novel cage molecules of type 1 is their ability to act as a superior catalyst for a continuously widening range of reactions[3-6] such as protecting alcohol groups with various silyl groups during multistep syntheses,[4] trimerizing isocyanates to isocyanurates[5] and the synthesis of alpha, beta-unsaturated nitriles.[6].
| Table 7. Verkade's bases |
|
55,695-5
|
2,8,9-Triisopropyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane
2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane,2,8,9-tris(1-methylethyl)]
C15H33N4P Mr 304.3 [175845-21-3] 1g, 5g
|
|
|
56,588-1
|
2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane
Purity ~97% C18H39N4P Mr 342.51 [331465-71-5] 1g, 5g
|
|
References:
- S. Arumugam, D. McLeod, J. G. Verkade, J. Org. Chem., 1997, 62, 3677.
- B. D'Sa, P. B. Kisanga, J. G. Verkade, J. Org. Chem., 1998, 63, 3691.
- P. B. Kisanga, B. D'Sa, J. G. Verkade, J. Org. Chem., 1998, 63, 10057.
- B. D'Sa, J. G. Verkade, "P(MeNCH2CH2)3N: An Efficient Silylation Catalyst," J. Am. Chem. Soc., 1996, 118, 12,832.
- J.-S. Tang, J. G. Verkade, "P(MeNCH2CH2)3N as a Superior Catalyst in the Conversion of Isocyanates to Isocyanurates," Angew. Chem., Int. Ed. Engl., 1993, 32, 896.
- P. B. Kisanga, J. G. Verkade, J. Org. Chem., 1999, 64, 4298.