Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
ChemFilesTitle Vol. 3, No. 2
Products For
Biomolecular NMR
Download PDF version (1.49 MB)
Page 1: Intro / Labeled Products for Cell Free Synthesis / Products for Peptide Synthesis / Products for Minimal Media
Page 2: Labeled Complex Growth Media / Buffers and Reagents / a-Keto Acids / Labeled Nucleotides
Page 3: Custom Synthesis / Aldrich Spectral Viewer / NEW - ISOTEC 2003-2004 Capabilities Catalog / ISOTEC Quality
Custom Synthesis
ISOTEC™ has the most experienced team of stable isotope custom synthesis chemists in the industry, led by an impressive group of Ph.D.'s, each one an expert in his field. ISOTEC™ routinely engages in multiple step syntheses of complex labeled molecules including metabolites and steroids.
"Let us help you design your
stable isotope labeled molecule!"
Please contact C.T. Tan, Ph.D. at: isosales@sial.com
CT_Tan
Top
Aldrich® Spectral Viewer™ FT-IR and FT-NMR Libraries
The Aldrich Spectral Viewer is an electronic reference book on CD-ROM that contains spectra from the Aldrich FT-IR and FT-NMR libraries. This easy-to-use software program is more powerful than a printed book, allowing text and data field searching, and the manipulation, printing, and exporting of spectra.The libraries may be used individually or may be combined together. The full set of libraries includes FT-IR spectra for over 11,000 compounds and FT-NMR spectra (both 300 MHz 1H and 75 MHz 13C spectra) for over 15,000 compounds.
spec-view1   spec-view2   spec-view3
Features:
  • Displays FT-IR spectra or FT-NMR spectra with chemical structure, and technical data.
  • Text searching by Chemical Name, Aldrich Catalog Number, CAS Number, and Molecular Formula.
  • Advanced data field searching on Chemical Category and physical properties such as Boiling Point, Melting Point, Density, Flash Point, Molecular Weight, and Refractive Index.
  • Spectra processing using Zoom, Peak Integration (NMR only), and Peak Labeling.
  • Print spectra, structure, technical data, integral table, and peak listings.
  • Export spectra to a file (bitmap, metafile, or JPEG formats) or to the clipboard (bitmap or metafile formats).
  • Web link button provides additional technical data from the Sigma-Aldrich website.
  • Expandable - As new spectral libraries become available, they can be added at a minimal fee with no subscription or maintenance fees required.
Version FT-IR 2002 library
(11,000 compounds)
FT-NMR standard library
 (11,800 compounds)
FT-NMR 2001
supplemental library
 (3,500 compounds)
Single user, commercial Z54,016-1 Z54,126-5 Z53,808-6
Single user, academic Z54,028-5 Z54,138-9 Z53,818-3
Network Z54,039-0 Z54,149-4 Z53,797-7
Demo program Z54,159-1    
To request a free demo or for more information, vist our website at
www.sigma-aldrich.com/spectralviewer, or send us an email at labequipment@sial.com
Top
Now available...  ISOTEC's 2003-2004 Capabilities Catalog
ISOTEC_Catalog3
Click here to request your FREE copy!
More than 2,000 products • Over 300 new additions
  • NEW products for nutrition and metabolic studies
  • NEW products for biomolecular NMR applications
  • NEW Product Application section
    • Structure Determination of
    • Biomolecules by NMR
    • Magnetic Resonance Imaging &
      Magnetic Resonance
    • Spectroscopy
    • Breath Tests
    • Nutrition & Metabolic Studies
    • Cyclotron Target Materials
    • Agriculture
    • Custom Synthesis
Let us help design your molecule!
ISOTEC has custom synthesis expertise unmatched by any other Lab.
For competitive prices, high quality products and services
contact ISOTEC™ 1-800-448-9760 or (937) 859-1808.
Top
ISOTEC_header
pyramid
Quality
  • Highest Isotopic Enrichment
  • Excellent Chemical Purity
  • Highest Quality Control Specifications
Innovation
  • Let us help design your molecule
  • Custom synthesis expertise
  • Continual refining of process
  • Continual New Product Development
Convenience
  • One Stop Shop
  • Product Bundling with other divisions of Sigma-Aldrich
  • Sigma-Aldrich offices in over 33 countries
  • We offer labeled (Isotec™) and unlabeled (Fluka) products for Biomolecular NMR
  • On location stock rooms (Stock Option)
  • Same day delivery of items in distribution centers
We are committed to the success of our Customers, Employees and Shareholders through leadership in Life Science, High Technology and Service.
BioFlo 3000® is a trademark of New Brunswick Scientific.
ISOGRO™, ISOTEC™, and Spectral Viewer™ are trademarks and Aldrich® is a registered trademark of Sigma-Aldrich Biotechnology, L.P.
Millipore® is a registered trademark of Millipore Corporation.
Novagen® is a registered trademark of CN Biosciences INC (CNBI).
Rosetta™ is a trademark of CN Biosciences INC (CNBI).
Visit our Web site at www.sigma-aldrich.com/Isotec to see the latest products for Biomolecular NMR or contact your local Sigma-Aldrich office