Aldrich® Chemistry Webinars
MIDA Boronate Building Blocks Webinar
Host
Martin D. Burke
Martin D. Burke, Ph.D.
Assistant Professor of Chemistry
Unversity of Illinois
at Urbana-Champaign, U.S.A.
Towards a General Platform for Small-Molecule Synthesis
This Aldrich Chemistry Webinar discusses the many physical and chemical properties of MIDA boronates that underlie their widespread utility, including:
  • Ease of preparation, purification, storage, and minimized environmental impact
  • Reversibly attenuated boronic acid reactivity that enables small-molecule synthesis via iterative cross-coupling
  • Capacity for slow-release cross-coupling that transforms even notoriously unstable 2-heterocyclic, vinyl, and cyclopropyl subunits into air-stable and highly effective cross-coupling partners
  • Compatibility with a wide range of common reaction conditions, which enables the multistep synthesis of structurally complex boronic acids from simple B-containing starting materials
For more information on MIDA boronates, please visit aldrich.com/mida.
Watch recorded webinar now (June 16, 2010)