Aldrich Chemistry Webinars
Aziridine Aldehydes as Reagents for Rapid and Chemoselective Synthesis of Complex Molecules
email this page to a friendProfessor A. K. Yudin
 Professor A. K. Yudin
 Chemistry Department
 University of Toronto
 Canada
 Watch Webinar Now!
Overview: By taking advantage of orthogonally reactive functional groups, amphoteric molecules deliver more efficient syntheses and enable novel chemical transformations. The amphoteric aziridine aldehyde reagents recently developed in our lab have become useful tools in reductive amination, aza-Michael reactions, amino allene and alkynyl amine synthesis, peptidomimetic ligation, as well as one-step access to complex alkaloid frameworks.
Of particular significance is the facility with which amphoteric reagents stitch together complex macrocyclic peptides and other ring structures out of linear precursors. A wide range of macrocyclic structures can be made in a one-step process mediated by amphoteric aziridine aldehydes. Control elements such as fluorescent tags, solubilizing groups, and conformation-tuning substituents are amenable to incorporation into cyclic peptide frameworks at a late stage of synthesis. This webinar will showcase our recent advances in this field.
 Webnar 2011
Areas covered in the webinar:
  • Discovery of unprotected aziridine aldehydes
  • Selected properties of aziridine aldehydes
  • Application of aziridine aldehydes in stereocontrolled construction of heterocycles
  • Application of aziridine aldehydes in linear peptidomimetic ligation
  • Application of aziridine aldehydes in macrocyclization reactions
  • Recommended procedures, pitfalls, and troubleshooting
Aziridine Aldehyde Reagents
707686   695513  695521 695556 
707686
695513
695521
695556