ChemNews
May 2008
      Hot News  |  Latest ChemBlogs  |  New Literature  |   New Products  |  Product Highlights May 2008       
Welcome to ChemNews!
Our May ChemNews showcases over 40 brand NEW and innovative products for Catalysis and other new building blocks and reagents. In addition, we're featuring the latest literature for chemical synthesis available from Sigma-Aldrich, as well our latest product highlights — all easily accessed from one convenient location.
Make sure you subscribe to ChemNews — all ChemNews subscribers can download quarterly pdf and sdf files that contain all the new products added during the year.
Check out the categories to the right and click on the links to view the latest products in that category.
    New Products
ChemBlogs Button
New Lab Start-Up Program Button
Back to Top
Hot News
NEW! Cheminar 3.2
Formation of C–C Bonds via Catalytic Hydrogenation
New Cross-Coupling Application Portal
See our comprehensive portfolio of catalysts, reagents and reactants for cross-coupling compiled in a series of ChemFiles, Technology Highlights and ChemBlogs featuring examples of applications using these products.
Latest ChemBlogs
ortho-Bromo- and ortho-Iodophenylboronic Acids—Remarkable Organocatalysts for the Mild and Atom-Economical, Direct Amidation and Diels–Alder Reactions
Dennis Hall and co-workers have recently disclosed the remarkable ability of ortho-bromo- and ortho-iodophenylboronic acids, [2-BrC6H4B(OH)2] and [2-IC6H4B(OH)2], to catalyze the direct amidation of free carboxylic acids with amines and the Diels–Alder reaction of free alpha,beta-unsaturated carboxylic acids with simple dienes. The noteworthy features of this discovery are:
  • 2-BrC6H4B(OH)2 and 2-IC6H4B(OH)2 are better catalysts for the direct amidation than the hitherto most efficient catalyst for the reaction, namely 3,4,5 F3C6H2B(OH)2.
  • Both reactions take place at room temperature in an atom-economical, byproduct-free fashion.
  • Both are operationally simple affording the pure products after a simple filtration and acid–base extractions (no chromatographic separations required).
  • The boronic acid catalyst is recovered from the basic aqueous phase in high yield.
  • The direct amidation reaction is broad in scope, and the Diels–Alder reaction remarkably selective.
  • The two reactions can be combined in an efficient, one-pot, two-step cascade sequence that is promoted by a single catalyst. Read More
ortho-Bromo- and ortho-Iodophenylboronic Acids - Remarkable Organocatalysts for the Mild and Atom-Economical, Direct Amidation and Diels-Alder Reactions
Back to Top
New Literature
Improved Catalysts and Ligands for Asymmetric Synthesis
Vol. 41, No. 1
Improved Catalysts
and Ligands for
Asymmetric Synthesis
(3.95 Mb PDF)
Asymmetric Catalysis Priveleged Ligands and Complexes
Vol. 8, No. 2
Asymmetric Catalysis
Priveleged Ligands and Complexes
(8.88 Mb PDF)
Chemical Ligation
Vol. 8, No. 1
Chemical Ligation
(2.94 Mb PDF)
New Products
NEW Palladium Catalysts
696676 Structure Image 697230 Structure Image 692921 Structure Image Homogeneous Palladium Catalysts Kit
696676 697230 692921 698911
Homogeneous Palladium
Catalysts Kit I
NEW Phosphines and Phosphine Precursors
697427 Structure Image
697427
697419 Structure Image
697419
692344 Structure Image
692344
692689 Structure Image
692689
695335 Structure Image
695335
694746 Structure Image
694746
694673 Structure Image
694673
694703 Structure Image
694703
Other NEW Catalysts
696668 Structure Image 685631 Structure Image 685046 Structure Image 696277 Structure Image
696668
685631
Mild coupling and cyclization catalyst.
685046
Useful catalyst for deuterium labeling.
696277
697575 Structure Image 696307 Structure Image 696250 Structure Image 696242 Structure Image
697575
696307
Multipurpose catalyst useful in reductions, carbene transfer reactions, aziridinations, and methyleneations.
696250
Efficient precatalysts for hydrosililation of carbonyl compounds
696242
Efficient precatalysts for hydrosililation of carbonyl compounds
NEW Alkynes and Allenes
694126 Structure Image
694126
694118 Structure Image
694118
664960 Structure Image
664960
696072 Structure Image
696072
683922 Structure Image
683922
669237 Structure Image
669237
694452 Structure Image
694452
NEW Boron Reagents for Suzuki Coupling
696579 Structure Image
696579
696587 Structure Image
696587
696595 Structure Image
696595
696552 Structure Image
696552
696609 Structure Image
696609
696544 Structure Image
696544
697400 Structure Image
697400
696560 Structure Image
696560
692638 Structure Image
692638
NEW Aldehydes
697915 Structure Image
697915
687170 Structure Image
687170
682489 Structure Image
682489
682454 Structure Image
682454
672238 Structure Image 696811 Structure Image 695556 Structure Image
672238
696811
695556
Stable 3-phenylaziridine-2-carboxaldehyde dimer; synthetic equivalent of the monomer
Back to Top
Technology Spotlights
Technology Spotlights
A-Phos Palladium Complexes for Efficient Suzuki Coupling
A-Phos Palladium Complexes for Efficient Suzuki Coupling image
Chiral Phosphoric Acids: New Organocatalysts for Reductive Amination Image
ChiroSolv Kits Image
Click Here for more product highlights.
Back to Top