Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Aldrich
Merrifield Peptide Resins
Technical Bulletin banner
Aldrich Logo
Number AL-142
Ion-Exchange Resins and
Related Polymeric Adsorbents
Aldrich Logo
Home | Amberlite® and Amberlyst® Resins | Dowex® Resins | Sephadex® Products
  Merrifield Peptide Resins | Polyethylenimine-Based Products | Polyamide Resins
IV. MERRIFIELD’S PEPTIDE RESINS
Rarely does research that is ultimately honored by a Nobel Prize result in specific products of particular use to the scientific community, as have these peptide resins, available from Aldrich since 1980.
These chloromethylated styrene/divinyl benzene copolymers have been crucial to the development of techniques and technology leading to the routine, convenient synthesis of peptides.
Table XIV
Catalog Number
Crosslinkage (%) Capacity
(meq Cl-/g)
Part. size
(mesh)
221481 1 ~1 200-400
221503 2 ~1 200-400
Home | Amberlite® and Amberlyst® Resins | Dowex® Resins | Sephadex® Products
  Merrifield Peptide Resins | Polyethylenimine-Based Products | Polyamide Resins