Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Aldrich
Sephadex Products
Technical Bulletin banner
Aldrich Logo
Number AL-142
Ion-Exchange Resins and
Related Polymeric Adsorbents
Aldrich Logo
Home | Amberlite® and Amberlyst® Resins | Dowex® Resins | Sephadex® Products
  Merrifield Peptide Resins | Polyethylenimine-Based Products | Polyamide Resins
III. SEPHADEX® PRODUCTS
These highly specialized gelfiltration and chromatographic media are composed of macroscopic beads synthetically derived from the polysaccharide, dextran. The organic chains are cross-linked to give a threedimensional network having functional ionic groups attached by ether linkages to glucose units of the polysaccharide chains.
Available forms include anion and cation exchangers, as well as gelfiltration resins, with varying degrees of porosity; bead sizes fall in discrete ranges between 20 and 300µ. See Tables XII and XIII.
Table XII
Catalog Number Description Functionality Bead size (µ)
Sephadex Ion-Exchange Resins
C25120 Sephadex-CM C-25 carboxymethyl 40-120
C50120 Sephadex-CM C-50 carboxymethyl 40-120
A25120 Sephadex-DEAE A-25 2-(diethylamino)ethyl 40-120
A50120 Sephadex-DEAE A-50 2-(diethylamino)ethyl 40-120
Q25120 Sephadex-QAE A-25 quaternary aminoethyl 40-120
Q50120 Sephadex-QAE A-50 quaternary aminoethyl 40-120
SPC25120 Sephadex-SP C-25 sulfopropyl 40-120
SPC50120 Sephadex-SP C-50 sulfopropyl 40-120
Table XIII
Catalog Number Description Bead size (µ)
Sephadex Gel-Filtration Resins
G10120 Sephadex G-10 40-120
G15120 Sephadex G-15 40-120
G2550 Sephadex G-25 20-50
G2580 Sephadex G-25 20-80
G25150 Sephadex G-25 50-150
G25300 Sephadex G-25 100-300
G5050 Sephadex G-50 20-50
G5080 Sephadex G-50 20-80
G50150 Sephadex G-50 50-150
G50300 Sephadex G-50 100-300
G7550 Sephadex G-75 20-50
G75120 Sephadex G-75 40-120
G10050 Sephadex G-100 20-50
G100120 Sephadex G-100 40-120
271217 Sephadex G-150 40-120
271233 Sephadex G-200 40-120
Home | Amberlite® and Amberlyst® Resins | Dowex® Resins | Sephadex® Products
  Merrifield Peptide Resins | Polyethylenimine-Based Products | Polyamide Resins