|
| Caution: |
|
Due to the hazardous nature of many of the products packaged in Aldrich UN-Pac containers we strongly recommend that all users read this bulletin carefully and completely before starting any actual laboratory/production work. If you are unsure of any of these procedures or need assistance, please contact us prior to use.
|
|
| UN-Pac cylinders |
|
The Aldrich UN-Pac packaging/dispensing system provides a convenient method for storing and dispensing laboratory and development scale quantities of high purity anhydrous solvents, fine organics, and other high hazard liquids. The properties of these products require the prevention of external contamination and/or personal exposure be kept to a minimum. Many of the liquids must be handled and stored without exposure to atmospheric moisture or oxygen. Fortunately, the design of the Aldrich UN-Pac system allows ready and convenient transfer of these liquids using the equipment and techniques described in detail in this bulletin. Aldrich provides the special equipment needed for handling these deposit containers and their contents. A complete and detailed listing of the equipment is described in the equipment selection of this bulletin.
|
UN-Pac-20-C 20L carbon steel Actual color is blue |
UN-Pac-100-C 100L carbon steel Actual color is blue |
|
chemists helping chemists in research and industry |
|
P.O. Box 355, Milwaukee, WI 53201 USA Telephone: 414-273-3850 • 800-558-9160 Fax: 414-273-4979 • 800-962-9591 TWX: 910-262-3052 Telex: 26 843 |
| This information is presented to assist you in evaluating our product. It is intended for use by technically-skiIled persons. We do not guarantee favorable results, and we assume no liability inconnection with its use. This information is not intended as a license to operate under, or arecommendation to infringe, any patent covering any material or use. Not for drug, food, cosmetic or household use. |
|