Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Aldrich
Equipment
Technical Bulletin banner
AL-208 (Europe only)
UN-Pac Packing System and Transfer Procedure for Type C
Introduction / SpecificationsRecommended Transfer Procedure / Equipment
Equipment:
We offer a wide variety of adapters and liquid-transfer apparatus. The liquid from all transfer lines exits through a stainless steel needle. Common syringe needles equipped with luer hubs can be used on some of supplied apparatus. However, due to the small gauge of available needles with luer hubs, unacceptably slow flow rate often results. Therefore, we offer transfer lines equipped with 1/8-inch ferrule locks for special 11-gauge needles to give a medium flow rate. In order to obtain a fast flow rate, we recommend using one of our transfer lines or needle-tubing connectors which is equipped with a 3/16-inch ferrule lock for a special 7-gauge needle.
Brass adapters can be reused many times and are inert to hydrocarbon and ether solvents. However, when corrosives are used or if acid is used in clean-up, then stainless steel adapters must be used. The valves on UN-Pac containers are brass and the cylinder is as the reagents shipped are not corrosive to brass stainless steel in the absence of atmospheric moisture.
Many customers may prefer to simply order a predesigned and fabricated transfer system. Therefore, we offer completely assembled outlet lines in brass and stainless steel. These are constructed of threaded pipe and metal-braided Teflon tubing and end in either a luer lock, 1/8-inch ferrule lock, or 3/16-inch ferrule lock for connection to a needle which must be ordered separately. The complete list of available transfer lines is given in Table 2 with a representative system shown in Figure 7. Technical Information Bulletin No. AL-150 gives a complete list of available needles for these transfer lines.
Table 2: Transfer lines for liquid removal from UN-Pac containers.
Pipe, valves*
flexible tubing
Needle
Connection**
Relative
flow rate
Catalog No.
BT Luer lock Slow Z150215
SST Luer lock Slow Z150223
BT 1/8-in FL Medium Z150231
SST 1/8-in FL Medium Z150258
BT 3/16-in FL Fast Z150266
SST 3/16-in FL Fast Z150274
BT-brass pipe, brass valves and brass-braided Teflon tubing
SST-stainless steel pipe, stainless steel valves, and stainless steel-braided Teflon tubing
FL-ferrule lock.
* Needle valve is for nitrogen flush of transfer line and requires a septum (Catalog No. Z100722 or Z124354) which must be ordered separately.
** Needle not provided; must be ordered separately.
Figure 7: Liquid-transfer, example Z150258
More information about adapters, tubing and equipment you can find in the following references.
  1. “Aldrich Kilo-Lab Cylinders Packing System and recommended Transfer Procedures.”;
    Aldrich Technical Information Bulletin No. AL-149
  2. “Equipment for Transfer of Liquids from Aldrich Kilo-Lab Cylinders.” ;
    Aldrich Technical Information Bulletin No. AL-150.
  3. “Handling Air- Sensitve Reagents”;
    Aldrich Technical Information Bulletin No. AL-134 (PDF, 176KB).
  4. “Equipment for Handling Air-Sensitive Reagents." ;
    Aldrich Technical Information Bulletin No. AL-135
  5. “The Aldrich Sure/Pac Cylinder Packaging System and Recommended Transfer Procedures.";
     Aldrich Technical Information Bulletin No. AL-136 (PDF, 168KB).
The foregoing discussions and procedures are given to assist our customers in the transfer and use of our products supplied in UN-Pac containers. The user must read this bulletin carefully, consult the figures, and follow each step in the transfer procedure. If you need further assistance at any time, please contact us.
chemists helping chemists in research and industry
P.O. Box 355, Milwaukee, WI 53201 USA Telephone: 414-273-3850 • 800-558-9160
Fax: 414-273-4979 • 800-962-9591 TWX: 910-262-3052 Telex: 26 843
This information is presented to assist you in evaluating our product. It is intended for use by technically-skiIled persons. We do not guarantee favorable results, and we assume no liability inconnection with its use. This information is not intended as a license to operate under, or arecommendation to infringe, any patent covering any material or use. Not for drug, food, cosmetic or household use.