|
Aldrich®
Appearance and Handling
 |
Number AL-118
Polybrene® Product No. 107689
(1,5-dimethyl-1,5-diazaundecamethylene polymethobromide, hexadimethrine bromide)
|
 |
|
Appearance:
|
|
Off-white crystals or powder
|
|
Handling:
|
|
Polybrene may cause irritation of the eyes, skin, and mucous membranes. Use in a chemical fume hood. Wear safety goggles and rubber gloves.
|
|
Emergency Procedures:
|
|
Fire May liberate acidic gases on burning. Extinguish with water spray, foam, carbon dioxide, or dry powder.
|
|
Spill Wear safety goggles and rubber gloves. Mix the material with dry caustic, sweep up, wrap in paper, and burn in an incinerator. Flush area well with water.
|
|
Skin Contact Wash with copious amounts of water. Remove contaminated clothing.
|
|
Eye Contact Wash with copious amounts of water. Consult a physician.
|
|
Ingestion Assume highly toxic if ingested. May cause lesions of the adrenal cortex. Induce vomiting. Consult a physician
|
|
Waste Disposal:
|
|
Mix the material with dry sodium bicarbonate, wrap in paper, and burn in an incinerator. Observe all Federal, state, and local regulations.
|
|
Storage:
|
|
Keep well closed. Extremely hygroscopic. Store in a cool dry place.