Sigma-Aldrich
Chemistry > Chemical Synthesis > Technology Spotlights > Asymmetric Allylic Alkylation
Chemical Synthesis
Asymmetric Allylic Alkylation
Introduction
Advantages
Pd-Catalyzed Asymmetric Allylic Alkylation:
   • Carbon Nucleophiles
   • Oxygen Nucleophiles
   • Nitrogen Nucleophiles
   • Sulfur Nucleophiles
Asymmetric Allylic Alkylation:
   • Molybdenum-catalyzed Reactions
Product Information
Headline graphic for Asymmetric Allylic Alkylation
Introduction
Palladium-catalyzed asymmetric allylic alkylation (AAA) has proven to be an exceptionally powerful method for the efficient construction of stereogenic centers. In sharp contrast to many other catalytic methods, AAA has the ability to form multiple types of bonds (C–C, C–O, C–S, C–N) with a single catalyst system.
C-2 symmetric diaminocyclohexyl (DACH) ligands in AAA
The Trost group at Stanford University has pioneered the use of C-2 symmetric diaminocyclohexyl (DACH) ligands in AAA, allowing for the rapid synthesis of a diverse range of chiral products with a limited number of chemical transformations. Reactions are typically high yielding, and excellent levels of enantioselectivity are observed.
Back to Top
DACH Table
References:
For comprehensive reviews, see:
  1. Trost, B. M.; Fandrick, D. R. Aldrichimica Acta 2007, 40, 59.
  2. Trost, B. M. et al. Acc. Chem. Res. 2006, 39, 747.
  3. Trost, B. M. J. Org. Chem. 2004, 69, 5813.
  4. Trost, B. M.; Crawley, M. L. Chem. Rev. 2003, 103, 2921.
  5. Trost, B. M.; Van Vranken, D. L. Chem. Rev. 1996, 96, 395.
  6. Trost, B. M. Acc. Chem. Res. 1996, 29, 355.
Advantages
  • Atom-economical catalytic method
  • High yields and synthetically useful levels of enantiocontrol
  • Unparallelled ability to prepare chiral building blocks from simple precursors
Back to Top
Representative Applications
Pd-Catalyzed Asymmetric Allylic Alkylation: Carbon Nucleophiles
In early examples of this methodology, Trost and co-workers demonstrated that diesters are competent nucleophiles for the deracemization of cyclic allylic acetates, to afford chiral malonate derivatives. Since that time, soft carbon nucleophiles such as barbituric acid derivatives, β-keto esters, nitro compounds, and many others have been employed in AAA for assembly of tertiary and quaternary asymmetric centers.
Malonate Nucleophiles:
Malonate Nucleophiles 1
References:
  1. Trost, B. M. Bunt, R. C. J. Am. Chem. Soc. 1994, 116, 4089.
  2. Ernst, M.; Helmchen, G. Synthesis 2002, 1953.
  3. Ernst, M.; Helmchen, G. Angew. Chem., Int. Ed. 2002, 41, 4054.
Malonate Nucleophiles 2
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 2005, 127, 14186.
Back to Top
Barbituric Acid Nucleophiles:
Barbituric Acid Nucleophiles
References:
  1. Trost, B. M.; Schroeder, G. M. J. Org. Chem. 2000, 65, 1569.
Back to Top
β-Keto Ester Nucleophiles:
ß-Keto Ester Nucleophiles
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 1997, 119, 7879.
Back to Top
Nitrosulfonyl Nucleophiles:
Nitrosulfonyl Nucleophiles 1
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 1998, 120, 1732.
Nitrosulfonyl Nucleophiles 1
References:
  1. Trost, B. M. et al. Chem.—Eur. J. 2001, 7, 3768.
Back to Top
Nitroalkane Nucleophiles:
Nitroalkane Nucleophiles 1
References:
  1. Trost, B. M.; Surivet, J. P. Angew. Chem., Int. Ed. 2000, 39, 3122.
Nitroalkane Nucleophiles 2
References:
  1. Trost, B. M.; Surivet, J. P. Angew. Chem., Int. Ed. 2000, 39, 3122.
Back to Top
Other Carbon Nucleophiles:
Other Carbon Nucleophiles 1
References:
  1. Trost, B. M.; Kallander, L. S. J. Org. Chem. 1999, 64, 5427.
Other Carbon Nucleophiles 2
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 2002, 124, 12420.
Other Carbon Nucleophiles 3
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 2005, 127, 2844.
  2. Trost, B. M. et al. J. Am. Chem. Soc. 2005, 127, 10259.
  3. Trost, B. M. et al. Chem.—Eur. J. 2005, 11, 951.
Back to Top
Pd-Catalyzed Asymmetric Allylic Alkylation: Oxygen Nucleophiles
Carbon-oxygen bond-forming reactions using palladium-catalyzed asymmetric allylic alkylation have been well demonstrated in numerous natural product syntheses. Alcohols, carboxylates, and hydrogencarbonates have all been employed as O-nucleophiles.
Alcohol Nucleophiles:
Alcohol Nucleophiles Structure
References:
  1. Lennon, I. C. et al. Chimica Oggi; Chemistry Today 2004, 11.
Alcohol Nucleophiles Structure
References:
  1. Fox, M. E. et al. Tetrahedron Lett. 2007, 48, 945.
Alcohol Nucleophiles 1
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 1998, 120, 12702.
Alcohol Nucleophiles 2
References:
  1. Trost, B. M. et al. Chem.—Eur. J. 2003, 9, 4442.
  2. Trost, B. M. et al. Org. Lett. 2000, 2, 4013.
  3. Trost, B. M.; Anderson, N. G. J. Am. Chem. Soc. 2002, 124, 14320.
  4. Trost, B. M.; Tang, W. Org. Lett. 2001, 3, 3409.
  5. Trost, B. M.; Zhang, T. Org. Lett. 2006, 8, 6007 (synthesis of allyl vinyl ethers).
Alcohol Nucleophiles 3
References:
  1.  
  2. Trost, B. M.; Toste, F. D. J. Am. Chem. Soc. 1999, 121, 3543.
  3. Trost, B. M.; Toste, F. D. J. Am. Chem. Soc. 2003, 125, 3090.
  4. Trost, B. M.; Crawley, M. L. J. Am. Chem. Soc. 2002, 124, 9328.
  5. Trost, B. M.; Crawley, M. L. Chem.—Eur. J. 2004, 10, 2237.
Back to Top
Carboxylate Nucleophiles:
Carboxylate Nucleophiles 1
References:
  1. Trost, B. M.; Organ, M. G. J. Am. Chem. Soc. 1994, 116, 10320.
Carboxylate Nucleophiles 2
References:
  1. Trost, B. M.; Kondo, Y. Tetrahedron Lett. 1991, 32, 1613.
Carboxylate Nucleophiles 3
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 1999, 121, 10834.
  2. Trost, B. M. et al. Chem.—Eur. J. 2001, 7, 1619.
Back to Top
Hydrogencarbonate Nucleophiles:
Hydrogencarbonate Nucleophiles 1
References:
  1. Lüssem, B. J.; Gais, H.-J. J. Am. Chem. Soc. 2003, 125, 6066.
  2. Gais, H.-J. et al. Tetrahedron Lett. 2005, 46, 6279.
Hydrogencarbonate Nucleophiles 2
References:
  1. Trost, B. M.; McEachern, E. J. J. Am. Chem. Soc. 1999, 121, 8649.
Hydrogencarbonate Nucleophiles 3
References:
  1. Gais, H.-J. et al. Tetrahedron Lett. 2005, 46, 6279.
  2. Dong, Y. et al. Tetrahedron Lett. 2005, 46, 353.
Back to Top
Pd-Catalyzed Asymmetric Allylic Alkylation: Nitrogen Nucleophiles
A formidable challenge in asymmetric synthesis is the stereocontrolled construction of carbon-nitrogen bonds. Nitrogen nucleophiles such as alkylamines, azides, amides, imides, and N-heterocycles have all been employed in asymmetric allylic alkylation reactions.
Alkylamines Nucleophiles:
Alkylamines Nucleophiles
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 1996, 118, 6297.
Back to Top
Azide Nucleophiles:
Azide Nucleophiles 1
References:
  1. Trost, B. M.; Pulley, S. R. J. Am. Chem. Soc. 1995, 117, 10143.
  2. Trost, B. M. et al. Chem.—Eur. J. 2001, 7, 1619.
Azide Nucleophiles 2
References:
  1. Trost, B. M.; Cook, G. C. Tetrahedron Lett. 1996, 37, 7485.
Back to Top
Sulfonamide Nucleophiles:
Sulfonamide Nucleophiles 1
References:
  1. Ovaa, H. et al. Chem. Commun. 2000, 1501.
  2. Trost, B. M.; Sorum, M. T. Org. Process Res. Dev. 2003, 7, 432.
Sulfonamide Nucleophiles 2
References:
  1. Stragies, R.; Blechert, S. J. Am. Chem. Soc. 2000, 122, 9584.
Back to Top
Imide Nucleophiles:
Imide Nucleophiles 1
References:
  1. Trost, B. M.; Van Vranken, D. L. J. Am. Chem. Soc. 1993, 115, 444.
  2. Trost, B. M. et al. J. Am. Chem. Soc. 1992, 114, 9327.
  3. Trost, B. M.; Patterson, D. E. J. Org. Chem. 1998, 63, 1339.
Imide Nucleophiles 2
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 1996, 118, 6520.
Imide Nucleophiles 3
References:
  1. Trost, B. M.; Bunt, R. C. J. Am. Chem. Soc. 1994, 116, 4089.
Imide Nucleophiles 4
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 2000, 122, 5968.
  2. Trost, B. M.; Lemoine, R. C. Tetrahedron Lett. 1996, 37, 9161.
  3. Trost, B. M. et al. Angew. Chem., Int. Ed. 2003, 42, 5987.
  4. Trost, B. M. et al. Chem.—Eur. J. 2006, 12, 6607.
Imide Nucleophiles 5
References:
  1. Trost, B. M.; Aponick, A. J. Am. Chem. Soc. 2006, 128, 3931.
Back to Top
Pd-Catalyzed Asymmetric Allylic Alkylation: Sulfur Nucleophiles
While not as extensively explored, certain sulfur nucleophiles are competent in the allylation reaction. In particular, sodium benzenesulfinate under goes alkylation to afford synthetically useful chiral sulfones.
Sulfur Nucleophiles 1
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 1995, 117, 9662.
Sulfur Nucleophiles 2
References:
  1. Trost, B. M. et al. J. Am. Chem. Soc. 2000, 122, 6120.
Back to Top
Asymmetric Allylic Alkylation: Molybdenum-catalyzed Reactions
The mechanism of molybdenum-catalyzed AAA reaction is presumed to be distinctly different from the analogous Pd-catalyzed reaction, and in some cases, levels regio-, enantio- and diastereoselectivity enhanced relative to the palladium-catalyzed reaction.
Asymmetric Allylic Alkylation: Molybdenum-catalyzed Reactions 1
References:
  1. Trost, B. M.; Zhang, Y. J. Am. Chem. Soc. 2006, 128, 4590.
Asymmetric Allylic Alkylation: Molybdenum-catalyzed Reactions 2
References:
  1. Trost, B. M.; Dogra, K. Org. Lett. 2007, 9, 861.
Back to Top
Product Information
Product # Product Name Structure Add to Cart
692794 (S,S)-DACH-phenyl Trost Ligand (S,S)-DACH-phenyl Trost Ligand
692808 (R,R)-DACH-phenyl Trost Ligand (R,R)-DACH-phenyl Trost Ligand
692786 (S,S)-DACH-naphthyl Trost Ligand (S,S)-DACH-naphthyl Trost Ligand
692778 (R,R)-DACH-naphthyl Trost Ligand (R,R)-DACH-naphthyl Trost Ligand
692743 (S,S)-DACH-pyridyl Trost Ligand (S,S)-DACH-pyridyl Trost Ligand
692751 (R,R)-DACH-pyridyl Trost Ligand (R,R)-DACH-pyridyl Trost Ligand
See these products in our Online Product Catalog