Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Technology Spotlights > BH3 THF Solutions
Chemical Synthesis
BH3 THF Solutions
As part of our ongoing commitment to safety, Sigma-Aldrich now introduces a NIMBA-stabilized BH3-THF solution. This solution has a practical advantage in that all pack sizes offered (800 mL, 1L, 8L, 18L, and bulk) meet Self-Accelerated Decomposition Temperature (SADT) thresholds required for safe shipment at room temperature, while demonstrating excellent reactivity and selectivity.
Sample Reactions
NIMBA_1
NIMBA_2
Click here for technical bulletin.
Ordering Information
Product Name Product # 
Borane-tetrahydrofuran complex, 1.0M solution in THF, stabilized with 0.005M N-isopropyl-N-methyl-tert-butylamine (NIMBA) new_icon21 650412
Borane-tetrahydrofuran complex, 1.0M solution in THF, stabilized with 0.005M Sodium Borohydride 176192