Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Drug Discovery
Product Highlights
Boc-Arylpiperazines
Privileged structures are a class of molecules that are capable of binding to multiple receptors with high affinity. In order to be considered privileged, a substructure should represent a molecule’s core element and make up a significant portion of its total mass. Recent reports1,2,3 have shown the presence of the arylpiperazine scaffold in potent and selective agonists of the human melanocortin-4 receptor (Fig 1).
Fig. 1
Sigma-Aldrich is proud to offer the following new BOC-protected arylpiperazines to our organic synthesis and drug discovery customers.
Product Name Structure Product #
1-BOC-4-(2-formylphenyl)piperazine, 97%
2-FORMYLPHENYL
65,151-6
1-BOC-4-(4-formylphenyl)piperazine, 97%
4-FORMYLPHENYL
65,142-7
1-BOC-4-(2-methoxycarbonylphenyl)piperazine
2-METHOXYCARBONYLPHENYL
65,237-7
1-BOC-4-(4-methoxycarbonylphenyl)piperazine, 97%
4-METHOXYCARBONYLPHENYL
65,238-5
1-BOC-4-(3-(ethoxycarbonyl)phenyl)piperazine, 97%
3-(ETHOXYCARBONYL)PHENYL
65,412-4
4-BOC-1-(5-bromo-2-pyridyl)piperazine, 97%
5-BROMO-2-PYRIDYL
65,376-4
4-BOC-1-(6-bromo-2-pyridyl)piperazine, 97%
6-BROMO-2-PYRIDYL
65,377-2
4-BOC-1-(6-methyl-2-pyridyl)piperazine, 97%
6-METHYL-2-PYRIDYL
65,375-6
4-(BOC-piperazin-1-yl)-3-nitrobenzoic acid, 97%
4-(BOC-PIPERAZIN-1-YL)
65,192-3
Methyl 2-(BOC-piperazin-1-yl)-5-nitrobenzoate, 97%
METHYL 2-(BOC-PIPERAZIN-1-YL)
65,193-1
References:
  1. Richardson, T. I. et al. J. Med. Chem. 2004, 47, 744.
  2. Pontillo, J. et al. Bioorg. Med. Chem. Lett. 2004, 14, 4417.
  3. Dyck, B. et al. Bioorg. Med. Chem. Lett. 2003, 13, 3793.