Chemistry > Chemical Synthesis > Technology Spotlights > Buchwald Precatalysts and Ligand Guide
Buchwald Precatalysts and Ligands
Dialkylbiaryl phospine ligands, and the precatalysts derived from them, are commonly referred to as Buchwald Precatalysts and Ligands. These reagents have developed into a highly valuable class of compounds for palladium catalysis and can now be used for a broad range of reactions. This brochure is designed to show the common uses and advantages of these powerful, and now commercially available, Buchwald Precatalysts and Ligands.
Buchwald Precatalysts
Buchwald Precatalysts are highly active for a range of Pd-catalyzed reactions. These bench-stable complexes can be activated under mildly basic conditions and provide a number of unique advantages over traditional palladium catalysts in terms of both activity and stability.
Figure 1
Figure 1. Different Pd Sources and Different Methods of Activation in C-N Cross-Coupling Reactions
Buchwald Ligands
Figure 2
Figure 2. Structural Features of Dialkylbiaryl Phosphine Ligands
Precatalysts
Product Structure Common Name Application for Pd Catalysis
704954 XPhos Palladacycle
  • α-Arylation of aldehydes
  • Buchwald-Hartwig amination
708739 tBuXPhos Palladacycle
  • α-Arylation of acetate esters
  • Buchwald-Hartwig amination
704946 SPhos Palladacycle
  • Buchwald-Hartwig amination
707589 RuPhos Palladacycle
  • Buchwald–Hartwig amination
718750 BrettPhos Palladacycle
  • Buchwald–Hartwig amination
741825
New
2nd Generation XPhos Palladacycle
  • Suzuki coupling
  • Low temperature Buchwald–Hartwig amination
753009
New
2nd Generation SPhos Palladacycle
  • Low temperature Buchwald–Hartwig amination
753246
Coming Soon
2nd Generation RuPhos Palladacycle
  • Low temperature Buchwald–Hartwig amination
Ligands
Product Structure Common Name Application for Pd Catalysis
638099 Cyclohexyl JohnPhos
  • Suzuki coupling
  • Negishi coupling
  • Hiyama coupling
  • Methylation of aryl halides
  • α-Arylation of ketones
  • Buchwald-Hartwig amination
  • C-B bond formation
  • Oxidation of alcohols to ketones
638021 DavePhos
  • Suzuki coupling
  • Kumada-Corriu coupling
  • α-Arylation of ketones
  • Buchwald-Hartwig amination
  • Rh catalyzed hydroamination
638064 XPhos
  • Suzuki coupling
  • Sonogashira coupling
  • Hiyama coupling
  • Stille coupling
  • Carbonyl enolate coupling
  • Buchwald-Hartwig amination
638072 SPhos
  • Suzuki coupling
  • Kumada-Corriu coupling
  • Negishi coupling
  • Buchwald-Hartwig amination
695262 MePhos
  • Suzuki coupling
  • α-Arylation of ketones
  • Buchwald-Hartwig amination
663131 RuPhos
  • Suzuki coupling
  • Negishi coupling
  • α-Arylation reaction of oxindoles
  • Buchwald-Hartwig amination
718742 BrettPhos
  • Buchwald-Hartwig amination
  • Trifluoromethylation of aryl chlorides
677280 SSPhos
  • Suzuki coupling
  • Rh-catalyzed 1, 2-addition of arylboronic acids to aldehydes and ketones
695882 PhDavePhos
  • α-Arylation of α-amino acid esters
  • Heteroarene benzylation
  • Buchwald-Hartwig amination
638080 tBuXPhos
  • α-Arylation of tetramic acids
  • Decarboxylative coupling of aromatic acids and aryl iodides
  • Carboxylation of aryl bromides with CO2
  • Buchwald-Hartwig amination
  • Buchwald-Hartwig C-O coupling
638439 JohnPhos
  • Suzuki coupling
  • Heck coupling
  • Buchwald-Hartwig amination
  • Buchwald-Hartwig C-O coupling
  • C-Si bond formation
675938 Tetramethyl di-tBuXPhos
  • Coversion of aryl halides to phenols
  • Buchwald-Hartwig amination
  • Buchwald-Hartwig C-O coupling
695211 tBuMePhos
  • α-Arylation of ketones
  • Buchwald-Hartwig C-O coupling
  • Formate reduction of N-heterocyclic allylic acetates
730998 tBuBrettPhos
  • Buchwald-Hartwig amination
  • C-F bond formation
  • O-arylation of ethyl acetohydroximate
  • Conversion of aryl and vinyl triflates to bromides and chlorides
  • Conversion of aryl chlorides, triflates, and nonaflates to nitroaromatics
695874 tBuDavePhos
  • α-Arylation of esters
  • Buchwald-Hartwig C-O coupling
  • Coupling of ammonia with aryl halides
  • C-Si bond formation
731013 JackiePhos
  • N-arylation of secondary amides
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Download (1MB)