Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Technology Spotlights > Catalysis Screening Kits
Chemical Synthesis
Catalysis Screening Kits
Accelerate Methodology Discovery, Lead Generation, and Process Optimization
Aldrich’s new chemistry kits for catalysis screening offer the broadest range of technologies available in convenient packaging. Kits contain sets of catalysts, achiral or chiral ligands, and scavengers with varying steric and electronic properties that will accelerate your research discoveries. Kits offer several advantages:
  • Enable rapid screening of catalytic processes for optimization
  • Conveniently packaged in sampler format
  • Cost of kit less than the total cost of individual components
New! Kits Containing New Aldrich Products
Buchwald Ligands I
Utilized for aromatic carbon-heteroatom formation and Suzuki coupling:
Buchwald_ligands
659932-1KT 1-g bottles
NHC Ligands I
Sterically and electronically varied to generate metal-based catalysts with different properties:
NHC_ligands
662232-1KT 1-g bottles
BINOL Ligands I
BINOLs are privileged ligands that exhibit high levels of enantiocontrol in various catalytic transformations:
Binols_ligands
669113-1KT
QuadraPureTM Scavengers
These macro- and microporous resins effectively remove metal contamination during chemical processing from R&D to manufacturing scale:
Quadrapures
659428-1KT 5-g bottles
Other Catalysis kits from Sigma-Aldrich
Homogeneous Pd Catalysts I
PdCl2 · Pd(OAc)2 · Pd2(dba)3
· Pd(PPh3)2Cl2 · Pd(PPh3)4 · [Pd(allyl)Cl]2 · Pd(dppf)Cl2
659975-1KT 1-g bottles
Heterogeneous Pd Catalysts I
· Palladium on Barium · Palladium on Calcium Carbonate · Palladium on Alumina · Palladium on Activated Carbon · Palladium (Dry) on Activated Carbon · Palladium Hydroxide
659967-1KT 1-g bottles
Pd EnCatTM
Pd EnCatTM 30 · Pd EnCatTM 40 · Pd EnCatTM TPP30 · Pd EnCatTM TOTP30 · Pd EnCatTM BINAP30
666335-1KT 1-g bottles
Phosphine Ligands I
PPh3 · PCy3 · P(t-Bu)3H-BF4 · P(o-tolyl)3
· P(2-Furyl)3 · dppp · dppf
659940-1KT 1-g bottles