Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Technology Spotlights > Click Chemistry
Chemical Synthesis
Click Chemistry
A "Click" Away From Discovery:
The traditional process of drug discovery based on natural secondary metabolites has often been slow, costly, and labor-intensive. Even with the advent of combinatorial chemistry and high-throughput screening in the past two decades, the generation of leads is dependent on the reliability of the individual reactions to construct the new molecular framework.
Click chemistry is a newer approach to the synthesis of drug-like molecules that can accelerate the drug discovery process by utilizing a few practical and reliable reactions. Sharpless and coworkers defined what makes a click reaction as one that is wide in scope and easy to perform, uses only readily available reagents, and is insensitive to oxygen and water. In fact, in several instances water is the ideal reaction solvent, providing the best yields and highest rates. Reaction work-up and purification uses benign solvents and avoids chromatography.1
Of the reactions comprising the click universe, the “perfect” example is the Huisgen 1,3-dipolar cycloaddition of alkynes to azides to form 1,4-disubsituted-1,2,3-triazoles (Scheme 1). The copper(I)-catalyzed reaction is mild and very efficient, requiring no protecting groups, and requiring no purification in many cases.2The azide and alkyne functional groups are largely inert towards biological molecules and aqueous environments, which allows the use of the Huisgen 1,3-dipolar cycloaddition in target guided synthesis3 and activity-based protein profiling.4 The triazole has similarities to the ubiquitous amide moiety found in nature, but unlike amides, is not susceptible to cleavage. Additionally, they are nearly impossible to oxidize or reduce.
Click chemistry Scheme 1 Image
Using Cu(II) salts with ascorbate has been the method of choice for preparative synthesis of 1,2,3-triazoles, but is problematic in biocojugation applications. However, tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, TBTA (Figure 1), has been shown to effectively enhance the copper-catalyzed cycloaddition without damaging biological scaffolds.5
Click chemistry Figure 1 Image
Recently, Sharpless and coworkers reported the ruthenium-catalyzed cycloaddition of azides to alkynes to form the complementary 1,5-disubstituted triazoles.6 Several ruthenium complexes were employed, but the pentamethylcyclopentadienyl (Cp*) analogues gave the best results, with Cp*RuCl(PPh3)2 being employed in most cases. Whereas the Cu(I)-catalyzed reaction is limited to terminal alkynes, the Ru(II)-catalyzed reaction is active with internal alkynes as well (Scheme 2).
Click chemistry Scheme 2 Image
Of course many aliphatic azides are not commercially available. Carreira and coworkers recently reported the hydroazidation of unactivated olefins to yield alkyl azides in the presence of a cobalt catalyst prepared in situ from a Schiff base ligand and Co(BF4)2·6H2O (Scheme 3).7 Additionally, the reaction can be coupled to the Sharpless cycloaddition to yield the 1,4-triazole in a one-pot process.
Click chemistry Scheme 3 Image
Sigma-Aldrich is pleased to offer these reagents and substrates for your research requirements in the exciting field of click chemistry.
Product Information
Product #  Product Name Add to Cart
Click Catalyst and Ligands
C1297 Copper(II) sulfate, ReagentPlus®, ≥99%
209198 Copper(II) sulfate pentahydrate ACS reagent, ≥98.0%
326755 Copper(II) acetate, 98%
205540 Copper(I) iodide, 98%
212865 Copper(I) bromide, 98%
A7631 (+)-Sodium L-ascorbate crystalline, ≥98%
673293 Pentamethylcyclopentadienylbis(triphenylphosphine)ruthenium(II) chloride
678937 tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, TBTA
667234 Chloro(pentamethylcyclopentadienyl)(cyclooctadiene)ruthenium(II)
Hydroazidation of Olefins
676551 Potassium 2-(3,5-di-tert-butyl-2-hydroxybenzylideneamino)-2,2-diphenylacetate, 95%
399957 Cobalt(II) tetrafluoroborate hexahydrate, 99%
Azide Sources
480525 Lithium azide solution, 20 wt. % in H2O 
438456 Sodium azide, 99.99+ % 
155071 Azidotrimethylsilane, 95% 
178756 Diphenyl phosphoryl azide, 97% 
349488 Azidotrimethyltin(IV), 97% 
651664 Tetrabutylammonium azide, 97% 
PEG Azides
17758 11-Azido-3,6,9-trioxaundecan-1-amine, technical, ≥90% (GC)
76172 O-(2-Aminoethyl)-O′-(2-azidoethyl)pentaethylene glycol, ≥90% (oligomer purity)
76318 O-(2-Aminoethyl)-O′-(2-azidoethyl)heptaethylene glycol, ≥90% (oligomer purity)
77787 O-(2-Aminoethyl)-O′-(2-azidoethyl)nonaethylene glycol , ≥90% (oligomer purity)
71613 O-(2-Azidoethyl)-O-[2-(diglycolyl-amino)ethyl]heptaethylene glycol, ≥90% (oligomer purity)
Organic Azides
59955 α-Azidoisobutyric acid solution, ~15% in heptane (T)
52916 α-Azidoisobutyric acid solution, purum, ~15% in heptane (T)
77213 Fluka  Ethyl azidoacetate solution, purum, ~25% in toluene (NMR) 
93528 Ethyl azidoacetate solution, purum, ~25% in ethanol (NMR)
244546 Azidomethyl phenyl sulfide, 95%
359556 4-Azidoaniline hydrochloride, 97%
359564 4-Azidophenyl isothiocyanate, 97%
276219 1-Azidoadamantane, 97%
514004 1-Azido-1-deoxy-β-D-glucopyranoside
513989 1-Azido-1-deoxy-β-D-galactopyranoside, 97%
M6691 α-D-Mannopyranosyl azide
152854 4-Methoxybenzyloxycarbonyl azide, 95%
340138 4-Carboxybenzenesulfonazide, 97%
A6057 4-Azidophenacyl bromide
11550 4-Azidophenacyl bromide, BioChemika, ≥98.0% (HPLC)
404764 4-Acetamidobenzenesulfonyl azide, 97%
A4810 3′-Azido-2′,3′-dideoxyuridine, ≥98% (TLC)
A2169 3′-Azido-3′-deoxythymidine, ≥98% (HPLC)
11546 3′-Azido-3′-deoxythymidine, BioChemika, ≥99.0% (HPLC)
11544 2′-Azido-2′-deoxyuridine, BioChemika, ≥99.0% (N) 
573213 (4S)-4-[(1R)-2-Azido-1-(benzyloxy)ethyl]-2,2-dimethyl-1,3-dioxolane
31755 7-(Diethylamino)coumarin-3-carbonyl azide, BioChemika, for fluorescence, ≥95% (HPLC)
493406 [3aS-(3aα,4α,5β,7aα)]-5-Azido-7-bromo-3a,4,5,7a-tetrahydro-2,2-dimethyl-1,3-benzodioxol-4-ol, 99%
A0456 (2S,3R,4E)-2-Azido-4-octadecene-1,3-diol
R109 Ro 15-4513
514012 1-Azido-1-deoxy-β-D-lactopyranoside  
A1262 8-Azidoadenosine 3′:5′-cyclic monophosphate, ~95%
283029 2,6-Bis(4-azidobenzylidene)-4-methylcyclohexanone, 97%
513970 1-Azido-1-deoxy-β-D-galactopyranoside tetraacetate, 97%
513997 1-Azido-1-deoxy-β-D-glucopyranoside tetraacetate
G4168 α-D-Mannopyranosyl azide tetraacetate, ≥90% (TLC)
A3407 6-(4-Azido-2-nitrophenylamino)hexanoic acid N-hydroxysuccinimide ester
E2028 Ethidium bromide monoazide, ≥95% (HPLC)
363227 4,4'-Diazido-2,2'-stilbenedisulfonic acid disodium salt hydrate
A6830 8-Azido-cyclic adenosine diphosphate-ribose, ≥95% (HPLC), lyophilized powder
56385 Photobiotin acetate, BioChemika, puriss., ≥98.0% (TLC)
back to the top
References:
  1. For recent reviews, see: (a) Kolb, H. C.; Sharpless, K. B. Drug Discovery Today 2003, 8, 1128. (b)Kolb, H. C. et al. Angew. Chem. Int. Ed. 2001, 40, 2004.
  2. (a) Rostovtsev, V. V. et al. Angew. Chem. Int. Ed.2002, 41, 2596.  (b) Tornøe, C. W. et al. J. Org. Chem. 2002, 67, 3057.
  3. (a) Manetsch, R. et al. J. Am. Chem. Soc.2004, 126, 12809. (b) Lewis, W. G. et al.Angew. Chem. Int. Ed. 2002, 41, 1053.
  4. Speers, A. E. J. Am. Chem. Soc. 2003, 125, 4686.
  5. Chan, T.R. et al. Org. Lett 2004,6, 2853.
  6. Zhang, L. et al. J. Am. Chem. Soc. 2005,127, 15998.
  7. Waser, J. et al. J. Am. Chem. Soc. 2005,127, 8294.