Sigma-Aldrich
Chemistry > Chemical Synthesis > Technology Spotlights > Gold Catalyst
Chemical Synthesis
Gold Catalyst
Introduction
Hashmi, Toste, Echavarren, and Haruta, among others, have fueled the advance of gold into the forefront of transition metal catalysis.1,2 Phosphine ligated gold(I) complexes have risen as powerful C–C, C–N, and C–O bond forming catalysts due to the ease with which they can activate C=C and C=C bonds thus allowing for unique rearrangements or reactions with various nucleophiles. Gold catalysis provides an excellent method to construct complex chemical architectures in a mild manner that would be difficult to achieve using other reaction paradigms. As illustrated below, the active catalysts are typically prepared by the addition of a silver activator to a gold halide precatalyst, although there are examples of isolable gold catalysts.
introduction
Sigma-Aldrich is proud to offer a treasure-trove of gold precatalysts and silver salts, as well as an extensive portfolio of unsaturated building blocks to accelerate your research success in this exciting field.
References: 
  1. For recent examples, see: (a) Shapiro, N. D.; Toste, F. D. J. Am. Chem. Soc., 2007,  129, 4160. (b) Jiménez-Núñez, E.; Echavarren, A. M. Chem. Commun. 2007, 333. (c) Echavarren, A. M. et al. Angew. Chem., Int. Ed. 2006, 45, 5452. (d) Haruta, M. Nature 2005, 437, 1098. (e) Luzung, M. R et al. J. Am. Chem. Soc. 2004, 126, 10858. (f) Hashmi, A. S. K. Gold. Bull. 2004, 37, 51. 
  2.  For recent reviews, see: (a) Hashmi, A. S. K.; Hutchings, G. J. Angew. Chem., Int. Ed. 2006, 45, 7896. (b) Widenhoefer, R. A.; Han, X. Eur. J. Org. Chem. 2006, 4555. (c) Zhang, L. et al. Adv. Synth. Catal. 2006, 348, 2271. (d) Hashmi, A. S. K. Angew. Chem., Int. Ed., 2005, 44, 6990.
  3. For a comprehesion review on gold catalysis, see: Hashmi, A. S. K. Chem. Rev. 2007, 107, 3180.
Advantages
  • Catalysts readily activate alkenes, alkynes, and alkenes in a myriad of transformations
  • Promote regio-, diastereo-, and enantioselective processes
  • Catalysts are typically air- and water-stable and reactions can be performed in an “open flask”
  • Exceptional functional group tolerance
  • Market exclusivity for selected gold catalysts
Representative Applications
Rearrangement of Allylic Acetates
Marion et al. reported the catalyzed isomerization of allylic acetates. Using a gold complex with a N-heterocyclic carbine ligand, the researchers reported the first gold catalyzed allylic rearrangement. To illustrate the versatility of the catalyst, several allyl acetates were rearranged with excellent yields.
Rearrangement of Allylic Acetates image
Reference:
Marion, N. et al. Org. Lett. 2007, 9, 2653.
Isomerization of Allenyl Carbinol Esters
Buzas et al. reported the synthesis of a series of 1,3-butadien-2-ol from different allenes using a gold complex as catalyst. Butadiene building blocks are of interest for Diels-Alder reactions, [4+1] cycloaddition and asymmetric hydrogenation. The catalyst utilized in this reaction is a gold complex with XPhos as a ligand. With a low loading of 1mol%, the authors were able to synthesize a variety of butadiene with yields up to 100%.
Isomerization of Allenyl Carbinol Esters image
Reference:
Buzas, A. K. et al. Org. Lett. 2007, 9, 985.
Intramolecular [4+2] Cycloadditions of Arylalkynes or 1,3-Enynes with Alkenes
Echavarren and co-workers reported the synthesis of pycnantuquinones A derivatives, catalyzed by [4+2] cycloaddition of dienynes. Using a stable crystalline gold complex based on the Buchwald ligand, the researchers were able to synthesize a family of derivatives with good yields.
Intramolecular [4+2] Cycloadditions of Arylalkynes or 1,3-Enynes with Alkenes image
Reference:
Nieto-Oberhuber, C. et al. J. Am. Chem. Soc. 2008, 130, 269.
Cyclopropanation
The Toste group demonstrated that olefins undergo stereoselective cyclopropanation (cis) with propargyl esters in the presence of in situ generated Ph3PAuSbF6 and is thus a complement to the trans selectivity observed in transition metal catalyzed cyclopropanation of olefins using α-diazoacetates.
scheme11s
Isomerization of 1,6-Enynes
Echavarren and others have studied a variety of gold catalyzed cyclizations of 1,6-enynes with or without the presence of nucleophiles. A variety of gold(I) catalysts are active, including Ph3PAuCH3, Ph3PAuSbF6, and Au[P(t-Bu)2(o-biphenyl)]SbF6 complexes. The reactions can tolerate heteroatoms located between the olefin and alkyne moieties.
Isomerization of 1,5-Enynes
Under gold(I) catalysis, 1,5-enynes of varying substitution patterns rearrange to give bicyclo[3.1.0]-hexenes in a high yielding, stereocontrolled fashion. For optically active substrates, the reaction can occur with efficient chirality transfer. The catalyst system utilizes Ph3PAuCl in combination with AgBF4, AgPF6, or AgSbF6 activators.
Isomerization of 1,4-Enynes (Rautenstrauch Rearrangement)
The Rautenstrauch rearrangement of 1,4-enynes provides an expeditious route to a diverse portfolio of functionalized cyclopentanones. Chiral 1-ethynyl-2-propenyl pivalates efficiently rearrange enantioselectively under mild conditions. Either Ph3PAuSbF6 or Ph3PAuOTf (both generated in situ) can be used, depending on the identity of the substrates.
Cyclization of ε-Acetylenic Carbonyls (Conia-Ene Reaction)
The non-catalyzed Conia-ene reaction provides access to methylenecyclopentanes without the need for deprotonation, however, the high temperatures required often result in diminished yields. Toste and co-workers reported a mild catalytic version of this reaction that proceeds under neutral conditions at ambient temperatures.
conia-ene_reaction
5-Endo-dig Carbocyclizations
While the gold(I)-catalyzed Conia-ene cycloisomerization is limited to terminal γ-alkynes, the related 5-endo-dig reaction allows for cyclization onto non-terminal δ-alkynes providing access to cyclopentene derivatives. This synthetic methodology can be applied to the preparation of simple bicyclic molecules, as well as in heterocycle synthesis (below) and halogenated cyclopentenes via alkynyl halide precursors.
5-Endo-dig_Carbocyclizations
Propargyl Claisen Rearrangement
The gold catalyst [(Ph3PAu)3O]BF4 provides access to a variety of homoallenic alcohols via a rapid two-step, one-pot sequence of a Claisen rearrangement of a propargyl vinyl ether followed by reduction of the aldehyde functionality. The reactions are generally high yielding and the robust catalyst system also shows superb ability to relay resident chirality into the allene products.
propargyl_claisen_rearrangement
Stereoselective Synthesis of Dihydropyrans
Using gold(I) catalysis, 2-substituted dihydropyrans are readily prepared from propargyl vinyl ethers in a stereocontrolled fashion.
synthesis_of_dihydropyrans
Hydroamination of Alkenes and Alkynes
Widenhoefer and He have explored a variety of gold catalyzed hydroamination reactions with alkenes and alkynes, and both intra- and intermolecular variants were studied. Using either Au(I) or Au(III) catalysts, amines, pyrrolidines, imines, and indoles can be easily accessed.
hydroamination
Hydrofunctionalization of Allenes with C, N, and O Nucleophiles
Vinylated tetrahydrofurans, tetrahydropyrans, pyrrolidines, and piperidines can be readily prepared from the corresponding heteroatom functionalized allenes using Au[P(t-Bu)2(o-biphenyl)]Cl and one of several silver activators. Alternatively, allene tethered indoles can be used in the preparation of carbazole derivatives.
hydrofunctionalization_of_allenes
Stereoselective Synthesis of Functionalized Dihydrofurans
Gagosz and co-workers have prepared a variety of 2,5-dihydrofurans via formation of an allene intermediate followed by cycloisomerization. The rapid reactions occur in the presence of Ph3PAuNTf2, and complete chirality transfer is observed.
functionalized_dihydrofurans
Ring Expansions of Alkynylcycloalkanols
1-Alkynylcycloalkanols rapidly rearrange to the corresponding 2-alkylidenecycloalkanones in the presence of several gold catalysts. The high yielding reactions provide a single olefin isomer when internal alkynes are employed.
ring_expansions
Acetylenic Schmidt Reaction
In the presence of a gold catalyst, pyrroles of varying substitution patterns can be prepared by an intramolecular acetylenic Schmidt reaction of homopropargyl azides.
acetylenic_schmidt_reaction
Product Information
Product Product Name Structure Add to Cart
404217
Chloro(trimethylphosphine)gold(I), 99%
404217
288225 Chloro(triethylphosphine)gold(I), 97% 288225
254037 Chloro(triphenylphosphine)gold(I), 99.9+% 254037
677922
Bis(trifluoromethanesulfonyl)imidate-(triphenylphosphinegold(I) 0.5 toluene adduct
677922
665177
Chloro(tris(4-trifluoromethylphenyl)phosphinegold(I), 99%
665177
679771
[1,1'-biphenyl-2-yl(di-tert-butyl)phosphine]chlorogold(I), 98%
679771
665185
Bis(diphenylphosphinomethane)dichlorodigold(I), 97%
665185
665142
Tris(triphenylphosphinegold)oxonium tetrafluoroborate
665142
677876
Trichloro(pyridine)gold(III), 97%
677876
696277
1,3-Bis(2,6-diisopropylphenyl-imidazol-2-ylidene)gold(I) chloride
1,3-Bis(2,6-diisopropylphenyl-imidazol-2-ylidene)gold(I) chloride
697575
(Acetonitrile)[(2-biphenyl)di-tert-butylphosphine]gold(I) hexafluoroantimonate
(Acetonitrile)[(2-biphenyl)di-tert-butylphosphine]gold(I) hexafluoroantimonate
695173
Dichloro(2-pyridinecarboxylato)gold
Dichloro(2-pyridinecarboxylato)gold
691674
2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl gold(I) bis(trifluoromethanesulfonyl)imide
2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl gold(I) bis(trifluoromethanesulfonyl)imide
687510
2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl gold(I) chloride
2-Dicyclohexylphosphino-2′,4′,6′-triisopropylbiphenyl gold(I) chloride
699616
Chloro[tris(2,4-di-tert-butylphenyl)phosphite]gold
Chloro[tris(2,4-di-tert-butylphenyl)phosphite]gold
481130
Gold(I) chloride, 99.9% (metals basis)
AuCl
379948
Gold(III) chloride, 99.99+%
AuCl3
G4022
Gold(III) chloride trihydrate, ≥49.0% as Au
HAuCl4 • 3H2O
298174
Sodium tetrachloroaurate(III) dihydrate, 99%
NaAuCl4 • 2H2O
398470
Gold(III) bromide hydrate, 99.9%
AuBr4 • xH2O
674583
Silver perchlorate, 97%
AgClO4
176427
Silver p-toluenesulfonate, ≥99%
Ag(OSO2C6H4CH3)
176435
Silver trifluoromethanesulfonate, ≥99%
Ag(OSO2CF3)
550256
Silver methanesulfonate
Ag(OSO2CH3)
T62405
Silver trifluoroacetate, 98%
Ag(CO2CF3)
208361
Silver tetrafluoroborate, 98%
AgBF4
227722
Silver hexafluorophosphate, 98%
AgPF6
227730
Silver hexafluoroantimonate(V), 98%
AgSbF6
668001
Silver bis(trifluoromethanesulfonyl)imide, 97%
AgN(OSO2CF3)2