Chemistry > Chemical Synthesis > Technology Spotlights > Hydrosilylation Catalyst
Chemical Synthesis
Hydrosilylation Catalyst
[Cp*Ru(MeCN)3]PF6: A Highly Efficient Hydrosilylation Catalysts
Vinylsilanes are versatile organometallic reagents that participate in a variety of reaction paradigms such as Tamao–Fleming oxidation, olefin metathesis, Pd-catalyzed cross-coupling, protodesilylation, and cycloaddition. Of the available methods for preparation of vinylsilanes, the hydrosilylation of alkynes is the most direct and atom-economical approach (Scheme 1). A number of transition metal catalysts have been devised to execute these reactions in a regio- and stereocontrolled fashion (Figure 1).
scheme_1s
figure_1
Methods for hydrosilylation of terminal alkynes were developed some time ago for the preparation of cis- and trans-β-vinylsilanes. Classical Pt-catalysis (Speier’s1 and Karstedt’s2 catalysts), as well as Rh-based catalysis ([Rh(cod)2]BF4 3 and [RhCl(nbd)]2 4), remain powerful methods for synthesis of trans-β-vinylsilanes. Wilkinson’s catalyst was also demonstrated to yield the trans product in polar solvents, with the cis isomer predominating in non-polar media.5 Ru-based catalysts (e.g. [Ru(benzene)Cl2]2 or [Ru(p-cymene)Cl2]2) allow for access to cis-β-vinylsilanes.6 Under certain conditions, Grubbs’ 1st generation catalyst also gives cis products, although the stereo- and regioselectivity of the hydrosilylation is highly dependent on the alkyne, silane, and solvent.7 While there exists a wealth of methods for preparation of linear β-vinylsilanes, until recently there were no general methods for the preparation of 1,1-disubstituted α-vinylsilanes.8 Moreover, although selective intramolecular hydrosilylation of internal alkynes can be achieved,9 a selective intermolecular variant was virtually unknown.10 The Trost group at Stanford University developed a remarkably robust protocol for hydrosilylation of terminal acetylenes to give α-vinylsilanes, relying on the ruthenium(II) catalyst, [Cp*Ru(MeCN)3]PF6.11-14 This catalyst also provides a competent method for regioselective intra- and intermolecular hydrosilylation of internal alkynes, giving exclusively Z-trisubstituted alkenes.
Intermolecular Hydrosilylation: Terminal Alkynes
A diverse set of terminal alkynes underwent rapid and mild hydrosilylation in the presence of [Cp*Ru(MeCN)3]PF6 to give 1,1-disubstituted α-vinylsilanes in good to excellent yield, often with low catalyst loadings (Scheme 2). The reaction is tolerant to a wide range of functional groups including halogens, free alcohols, alkenes, internal alkynes, esters, and amines. Moreover, a breadth of silanes can be used in the reaction with excellent predictability.
scheme_2s
Intermolecular Hydrosilylation: Internal Alkynes
As illustrated in Scheme 1, non-selective hydrosilylation of internal alkynes would potentially give four isomeric addition products. The Trost group has demonstrated that hydrosilylation of internal alkynes with [Cp*Ru(MeCN)3]PF6 gives trisubstituted Z-vinylsilanes exclusively, as a result of trans addition of the silane to the alkyne (Scheme 3).11
scheme_3s
Importantly, the hydrosilylation reaction displays a high level of regioselectivity. The regioselectivity can be summarized as follows: (i) hydrosilylation of 2-alkynes results in the formation of Z-alkenes with the silyl group occupying the less sterically-demanding position (entries 1 and 2); (ii) for substrates where the alkyne is not in the 2-position, the silyl substituent will occupy the more sterically-demanding position in the Z-alkene (entry 4); (iii) for propargylic, homopropargylic, and bishomopropargylic alcohol substrates, hydrosilylation occurs such that the silyl group resides distal to the hydroxyl functionality of the Z-alkene (entries 5–9); (iv) in the case of α,β-alkynylcarbonyls, the silyl group again selectively occupies the distal position of the Z-alkene (entries 10–13).11,15 For free propargylic, homopropargylic, and bishomopropargylic alcohols, hydrosilylation with a silane bearing a leaving group (e.g., an ethoxy substituent) results in the formation of a cyclic siloxane (entries 5 and 8). Significantly, the hydrosilylation using [Cp*Ru(MeCN)3]PF6 can be performed while maintaining the stereochemical integrity of asymmetric centers residing in the alkyne substrate (entry 9). Lastly, although non-sterically differentiated alkynes undergo stereo but non-regioselective hydrosilylation (entry 3), protodesilylation of the product mixture provides a single trans diastereomer.13 This provides a valuable complement to the cis-selectivity observed under Lindlar reduction conditions.
As shown in Scheme 4, even highly reactive silanes can participate in intermolecular hydrosilylation reaction, with excellent predictability.11b The resultant alkenylchlorosilane was trapped with a hexadienol to give a siloxane linkage. Heating the triene resulted in an intramolecular Diels-Alder (IMDA) reaction, yielding a siloxane with four contiguous stereocenters. The adduct could then be treated to protodesilylation or Tamao–Fleming conditions to furnish the primary alcohol or the diol respectively.
scheme_4s
Manipulation of the alkene prior to protodesilylation or oxidation is also feasible. For example, vinylsilanes are readily epoxidized by m-CPBA in a diastereoselective fashion (Scheme 5). Subsequent protodesilylation furnishes the corresponding syn-epoxy alcohol, while Tamao–Fleming oxidation provides a syn-diol. Therefore, this process can be used as a surrogate to the aldol condensation.
scheme_5s
Intramolecular Hydrosilylation
Finally, intramolecular hydrosilylation is possible using hydroxyalkynes, as illustrated in the concise synthesis of the 3-hydroxypiperidine alkaloid (+)-spectaline (Scheme 6).16b Treatment of the homopropargyl alcohol with tetramethyldisilazane (TMDS), followed by regio- (distal) and stereoselective (Z) intramolecular hydrosilylation, gave a cyclic azidosiloxane. Tamao–Fleming oxidation followed by reduction with concomitant cyclization gave (+)-spectaline in respectable yield.
scheme_6s
The sequence of intramolecular hydrosilylation and subsequent cross-coupling provides an excellent method for introducing a new carbon bond at an alkyne carbon that is in a remote position from a free hydroxyl group (Scheme 7).17
scheme_7
We are pleased to offer [Cp*Ru(MeCN)3]PF6, as well as a number of other catalysts for hydrosilylation.
   Product Information
Product # Product Name/Description Structure Add to Cart
667412 Pentamethylcyclopentadienyltris(acetonitrile)-ruthenium(II) hexafluorophosphate 667412
398322 Chloroplatinic acid hydrate H2PtCl6 · x H2O
479519 Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution, 0.10 M in xylene 479519
334987 Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate hydrate, 97% 334987
249939 (Bicyclo[2.2.1]hepta-2,5-diene)rhodium(I) chloride dimer 249939
199982 Tris(triphenylphosphine)rhodium(I) chloride 199982
341568 Benzenedichlororuthenium(II) dimer 341568
343706 Dichloro(p-cymene)ruthenium(II) dimer 343706
579726 Benzylidenebis(tricyclohexylphosphine)dichloro-ruthenium(II) 579276
References:
(1) For a recent example, see: Denmark, S. E.; Wehrli, D. Org. Lett. 2000, 2, 565.
(2) For recent examples, see: (a) Itami, K. et al. J. Org. Chem. 2002, 67, 2645. (b) Denmark, S. E.; Wang, Z. Org. Lett. 2001, 3, 1073. (c) Denmark, S. E.; Wang, Z. Org. Synth. 2005, 81, 54.
(3) Takeuchi, R. et al. J. Org. Chem. 1995, 60, 3045.
(4) Sato, A. et al. Org. Lett. 2004, 6, 2217.
(5) Takeuchi, R.; Tanouchi, N. J. Chem. Soc., Perkin Trans. 1 1994, 2909.
(6) Na, Y.; Chang, S. Org. Lett. 2000, 2, 1887.
(7) (a) Maifeld, S. V. et al. Tetrahedron Lett. 2005, 46, 105. (b) Menozzi, C. et al. J. Org. Chem. 2005, 70, 10717. (c) Aricó, C. S.; Cox, L. R. Org. Biomol. Chem. 2004, 2,, 2558.
(8) While references 7a and 7b illustrate access to α-vinylsilanes using Ph3SiH, the method does not appear to be general. Isomeric mixtures are often formed with other silanes.
(9) For recent examples, see: (a) Denmark, S. E.; Pan, W. Org. Lett. 2003, 5, 1119. (b) Denmark, S. E.; Pan, W. Org. Lett. 2002, 4, 4163. (c) Denmark, S. E.; Pan, W. Org. Lett. 2001, 3, 61.
(10) For a recent example, see: Hamze, A. et al. Org. Lett. 2005, 7, 5625.
(11) (a) Trost, B. M. et al. J. Am. Chem. Soc. 2001, 123, 12726. (b) Trost, B. M.; Ball, Z. T. J. Am. Chem. Soc. 2005, 127, 17644.
(12) For a mechanistic rationale of this Ru-catalyzed hydrosilylation, see: Chung, L. W. et al. J. Am. Chem. Soc. 2003, 125, 11578.
(13) Trost, B. M. et al. J. Am. Chem. Soc. 2002, 124, 7922.
(14) Trost, B. M. et al. Org. Lett. 2003, 5, 1895.
(15) Trost, B. M.; Ball, Z. T. J. Am. Chem. Soc. 2004, 126, 13942.
(16) (a) Trost, B. M. et al. Angew. Chem. Int. Ed. 2003, 42, 3415. (b) Trost, B. M. et al. J. Am. Chem. Soc. 2005, 127, 10028. (c) Trost, B. M. et al. Org. Lett. 2005, 7, 4911.
(17) Trost, B. M.; Ball, Z. T. J. Am. Chem. Soc. 2003,, 125, 30.