Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Technology Spotlights > Leighton's Strained Silacycles
Chemical Synthesis
Leighton's Strained Silacycles
Leighton’s Strained Silacycles: Powerful Enantioselective Allylation Reagents
    Enantioselective allylation of aldehydes
    Practical, enantioselective allylation of acylhydrazones
    Enantioselective allylation of ketimines: synthesis of tertiary carbinamines
    Friedel-Crafts Alkylation with Acylhydrazones
    Acylhydrazone-Alkene [3+2] Cycloaddition Reactions
    Products
The asymmetric allylation of carbonyl compounds remains one of the most important and fundamental addition reactions for the synthesis of optically active chiral building blocks. Prof. James L. Leighton from Columbia University has developed a powerful method making use of silicon in constrained five-membered ring systems benefiting of an increased reactivity. These five-membered silacycles consisting of allylsilanes and a simple chiral 1,2-diol, diamine or aminoalcohol showed profound practical implications acting as chiral silicon-based Lewis acids in allylation reactions.
back to top
Enantioselective allylation of aldehydes
In 2002, Leighton and co-workers developed strained silacycle compounds as versatile reagents for the practical enantioselective allylation of aldehydes.1
A newly developed chiral auxiliary (Aldrich Prod. No. 704725) based on the cyclohexane-1,2-diamine scaffold successfully allylated a broad range of aldehydes highly enantioselectively.2
Leighton Silacycles - 1
back to top
Practical, enantioselective allylation of acylhydrazones
The development of practical enantioselective syntheses of chiral amines is of great importance to synthetic organic and medicinal chemists. In 2003, Leighton and co-workers successfully used a pseudoephedrine-derived, five-membered-ring, strained silacycle reagent (Aldrich Prod. No. 706671) for the enantioselective allylation of acylhydrazones.3
Leighton Silacycles - 2
back to top
Enantioselective allylation of ketimines: synthesis of tertiary carbinamines
The reaction scope of these silacycles was extended to a practical method for the enantioselective synthesis of tertiary carbinamines based on the addition of this chiral allylsilane reagent to a structurally diverse array of ketone-derived benzoylhydrazones.4 While many methods for the synthesis of quaternary α-amino acids have been published, far fewer reports have dealt with the synthesis of tertiary carbinamines. The free amines are easily accessed in good yields by subjecting the different hydrazones to reduction with SmI2 (Aldrich Prod. No. 409340).
Leighton-Silacycles - 3
back to top
Friedel-Crafts Alkylation with Acylhydrazones
Addition of electron-rich arenes to glyoxylate-derived imines, in an overall Friedel-Crafts-like alkylation reaction, provides access to valuable arylglycine derivatives. Silacycle 720380 smoothly promotes the additions of anilines to the benzoylhydrazone of isopropyl glyoxylate.
Leighton-Silacycles - 4
back to top
Acylhydrazone-Alkene [3+2] Cycloaddition Reactions
A variety of aliphatic and aromatic aldehyde-derived benzoylhydrazones may be reacted with tert-butyl vinyl ether to give pyrazolidines with excellent levels of enantioselectivity. This reaction was easily carried out on a larger scale (5g of hydrazone) to demonstrate the true practicality and scalability of this method.
Leighton-Silacycles - 5
back to top
Products
Prod. No. Product Name Structure
704725 (R,R)-1,3-Bis[(4-bromophenyl)methyl]-2-chlorooctahydro-2-allyl-1H-1,3,2-benzodiazasilole  704725 Structure
705098 (S,S)-2-Allyl-1,3-bis-(4-bromobenzyl)-2-chlorooctahydro-2-1H-1,3,2-benzodiazasilole  705098 Structure
706671 (4S,5S)-2-Allyl-2-chloro-3,4-dimethyl-5-phenyl-1-oxa-3-aza-2-silacyclopentane 706671 Structure 
719056 (4R,5R)-2-Allyl-2-chloro-3,4-dimethyl-5-phenyl-1-oxa-3-aza-2-silacyclopentane 719056 Structure 
720380 (4S,5S)-2-Chloro-3,4-dimethyl-2,5-diphenyl-1-oxa-3-aza-2-silacyclopentane  720380 Structure
720380 (4R,5R)-2-Chloro-3,4-dimethyl-2,5-diphenyl-1-oxa-3-aza-2-silacyclopentane  720380 Structure
back to top
Reference:
  1. Review: Leighton, J. L. Aldrichimica Acta 2010, 31, 3-12.
  2. Kinnaird, J. W. A.; Ng, P. Y.; Kubota, K.; Wang, X., Leighton, J. L. J. Am. Chem. Soc. 2002, 124, 7920.
  3. Kubota, K.; Leighton, J. L. Angew. Chem. Int. Ed. 2003, 42, 946.
  4. Berger, R.; Rabbat, P. M. A.; Leighton, J. L. J. Am. Chem. Soc. 2003, 125, 9596.
  5. Shirakawa, S.; Berger, R.; Leighton, J. L. J. Am. Chem. Soc. 2005, 127, 2858.
  6. Shirakawa, S.; Lombardi, P. J.; Leighton, J. L. J. Am. Chem. Soc. 2005, 127, 9974.
back to top