Sigma-Aldrich
Chemistry > Chemical Synthesis > Technology Spotlights > MeOBIPHEP
Technology Spotlight
MeOBIPHEP
Introduction
State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design. In many respects, the catalytic profile of the MeOBIPHEP ligands is rather similar to that of other atropisomeric diphosphines such as BINAP and its many analogs.1–4 The nature of the PR2 group strongly influences the catalytic performance of the metal complexes, and the ligands available in research quantities from Sigma-Aldrich represent examples with different steric bulk.
References:
  1. Blaser, H.-U. et al. Chimica Oggi Supplement Catalytic Applications 2007, 25(2), 8.
  2. Blaser, H.-U. et al. Acc. Chem. Res. 2007, 40, 1240.
  3. Tang, W.; Zhang, X. Chem. Rev. 2003, 103, 3029.
  4. Schmid, R. et al. Pure Appl. Chem. 1996, 68, 131.
Representative Applications
Hydrogenation of α- and β-functionalized ketones
Ru-MeOBIPHEP complexes are highly effective for the hydrogenation of α- and β-functionalized ketones. Studied by Genêt and co-workers, a range of alkyl- and aryl β-keto esters and β-keto sulfones could be reduced in very high enantioselectivities using 29510 (1st eq.). A p-chlorophenylglyoxylic acid derivative could be successfully enantioselectively reduced on bench scale using 29510 to give the corresponding mandelic acid amide depicted with 93% ee and with a TON of ca. 4000 h-1 (2nd eq.). Genêt also described the synthesis of a chiral 2-chloro-1,3-dicarbonyl compound via dynamic kinetic resolution using Ru-MeOBIPHEP 29510 (3rd eq.). Finally, the Hamada group successfully applied Ir-MeOBIPHEP catalysts 29510 in the highly diastereo- and enantioselective reduction of β-hydroxy-α-amino esters (4th eq.).
MeO-BIPHEP Scheme 1 Image
Scheme 1
References
  1. Ratovelomanana-Vidal, V. et al. Adv. Synth. Catal. 2003, 345, 261.
  2. Blanc, D. et al. J. Organomet. Chem. 2000, 603, 128.
  3. Bertus, P. et al. Tetrahedron: Asymmetry 1999, 10, 1369.
  4. Cederbaum, F. et al. Adv. Synth. Catal. 2004, 346, 842.
  5. Mordant, C. et al. Synthesis 2003, 2405.
  6. Makino, K. et al. Org. Lett. 2006, 8, 4573.
back to top
Hydrogenation of allylic alcohols and α,β-unsaturated esters
MeOBIPHEP ligands are used in an impressive number of industrial hydrogenation processes on pilot scale developed by Roche. Not only enantioselectivities are excellent, but also TON and TOFs are on an impressively high level for various substrates.
MeOBIPHEP Scheme 2 Image
Scheme 2
References
  1. Schmid, R.; Scalone M. Comprehensive Asymmetric Catalysis; Jacobsen, E. N., Yamamoto, H., Pfaltz, A., Eds.; Springer: Berlin, 1999; p 1439.
  2. Crameri, Y. et al. Chimia 1997, 51, 303.
  3. Netscher, T. et al. Large Scale Asymmetric Catalysis; Blaser, H.-U., Schmidt, E., Eds.; Wiley-VCH: Weinheim, 2004; p 71.
back to top
Hydrogenation of heteroarenes
Wang et al. described the enantioselective hydrogenation of quinoline derivatives with an Ir-MeOBIPHEP 29510 catalyst. This method provides an efficient access to a variety of optically active tetrahydroquinolines with up to 96% ee. This methodology has been applied to the asymmetric synthesis of three naturally occurring alkaloids angustureine, galipinine, and cuspareine.
MeO-BIPHEP Scheme 3 Image
Scheme 3
Reference
Wang, W.-B. et al. J. Am. Chem. Soc. 2003, 125, 10536.
back to top
C–C coupling reactions
The Krische group reported on the enantioselective Rh-catalyzed reductive coupling of acetylene to aldehydes to α-ketoesters using 29510 to yield the corresponding diene alcohols in good yields and enantioselectivities (1st eq.). The Wiedenhoefer group reported on the intramolecular asymmetric hydroarylation of 2-(4-pentenyl)indoles in moderate to good yield with up to 90% ee catalyzed by a 1:1 mixture of dichloro platinum complex and (S)-3,5-t-Bu-4-MeO-MeOBIPHEP 29511 (2nd eq.). Pregosin and co-workers reported a dramatic improvement of enantioselection in the Pd-catalyzed Heck reaction of aryl triflates to 2-substituted dihydrofuranes when using (R)-3,5-t-Bu-MeOBIPHEP 29524 (3rd eq.).
MeOBIPHEP Scheme 4 Image
Scheme 4
References
  1. Kong, J. R.; Krische, M. J. J. Am. Chem. Soc. 2006, 128, 16040.
  2. Han, X.; Widenhoefer, R. A. Org. Lett. 2006, 8, 3801.
  3. Tschoerner, M. et al. Organometallics 1999, 18, 670.
back to top
Product No. Product Name Structure Add to Cart
29510 (R)-(+)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis(diphenylphosphine) 29510 Structure Image
29511 (S)-(-)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis(diphenylphosphine) 29511 Structure Image
29512 (R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphine] 29512 Structure Image
29513 (S)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphine] 29513 Structure Image
29514 (R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis(di-2-furylphosphine) 29514 Structure Image
29515 (S)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis(di-2-furylphosphine) 29515 Structure Image
29516 (R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(3,5-dimethylphenyl)phosphine] 29516 Structure Image
29517 (S)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(3,5-dimethylphenyl)phosphine] 29517 Structure Image
29518 (R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis(diisopropylphosphine) 29518 Structure Image
29519 (S)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis(diisopropylphosphine) 29519 Structure Image
29520 (R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(4-methylphenyl)phosphine] 29520 Structure Image
29521 (S)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(4-methylphenyl)phosphine] 29521 Structure Image
29524 (R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(3,5-di-tert-butylphenyl)phosphine] 29524 Structure Image
29525 (S)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(3,5-di-tert-butylphenyl)phosphine] 29525 Structure Image
29526 (R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(3,4,5-trimethoxyphenyl)phosphine] 29526 Structure Image
29527 (S)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis[bis(3,4,5-trimethoxyphenyl)phosphine] 29527 Structure Image
29528 (R)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis{bis[3,5-diisopropyl-4-(dimethylamino)phenyl]phosphine} 29528 Structure Image
29529 (S)-(6,6'-Dimethoxybiphenyl-2,2'-diyl)bis{bis[3,5-diisopropyl-4-(dimethylamino)phenyl]phosphine} 29529 Structure Image