Chemical Synthesis
Os EnCat 40
|
Os EnCatTM 40 Microencapsulated OsO4
Sigma-Aldrich is pleased to offer innovative catalysts for use in practical synthetic transformations in collaboration with Reaxa. Reaxa has immobilized osmium tetroxide, a toxic and volatile reagent, via encapsulation in a polyurea matrix to create Os EnCatTM 40 catalyst (cf. pictures below). This innovative product offers safety and handling advantages versus conventional OsO4, while acting as an alternative reagent in the industrially useful dihydroxylation reaction paradigm.
Product Ordering Information Os EnCat™40, 0.3 mmol/g Os loading: 658685
|
 Optical Image of Os EnCatTM40 Beads |
 Os EnCatTM40 and OsO4 visual after 2 months storage |
|
|
Advantages of Os EnCat™ 40 Catalyst:
· Safer, easier to handle, and non-volatile · Greater storage stability versus OsO4 · Facile recovery of catalyst · Low levels of Os metal in final product · Catalyst can be recycled
Reactivity of Os EnCat
Application Data
Os EnCatTM 40 has shown good catalytic activity in the dihydroxylations of acyclic and cyclic olefins of varying substitution patterns (Table I below). Catalyst efficiency is heightened by use of a single phase solvent in conjunction with the co-oxidant 4-methylmorpholine-N-oxide (NMO, 224286). Amounts of Os metal in the crude product mixture are normally in the 10-40 ppm range, which is acceptable for most pharmaceutical purity assays. To achieve optimum reaction rates, processes with Os EnCatTM are normally performed at elevated temperatures. Os EnCatTM 40 can be utilized in combination with chiral ligands such as [(DHQD)2PHAL] (Aldrich product 392766) to induce stereoselective transformations. Os EnCatTM 40 has also shown to be an effective catalyst for the oxidative cleavage of olefins to generate their respective carbonyl compounds under mild reaction conditions.
Dihydroxylation of Acyclic and Cyclic Olefins Using Os EnCatTM 40*:
 Table I. *Conditions: Olefin (1 mmol), N-methylmorpholine-N-oxide (1.5 mmol), solvent (10 mL).
|
| Product Name |
Product # |
Os EnCat™ 40: 0.3 mmol/g Os loading  |
658685 |
| 4-Methylmorpholine N-oxide 97% |
224286 |
| AD-mix-β |
392766 |