Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Technology Spotlights > Benzoylation Reagents
Chemical Synthesis
Benzoylation Reagents
Simple Tools for α-Oxygenation of Aldehydes and Ketones
The α-hydroxycarbonyl moiety is an important structural motif in organic synthesis and is present in a substantial number of natural products.  A variety of methods have been developed to prepare this functionality including α-oxygenation of enolates with electrophilic oxidizing agents as well as dihydroxylation or epoxidation of enol ethers.1 Recently, aminooxylation of aldehydes 2 and ketones3 has been demonstrated using nitrosobenzene.  However, this latter protocol requires a large excess of the carbonyl compound in tandem with syringe pump techniques which diminishes from the usefulness of the transformation.
Nicholas Tomkinson of Cardiff University (UK) recently reported several practical reagents for preparation of  N-tert-butyl-O-benzoylhydroxylamine hydrochloride (669407) is a bench stable reagent for α-functionalization of a variety of aldehydes under mild conditions (Scheme 1).  The reaction can be performed in open air and in the presence of moisture.  Benzoylation occurs in high yield at both secondary and tertiary centers as demonstrated by the reactions of this reagent with isovaleraldehyde and cyclohexanecarboxaldehye (Figure 1).   
scheme1
figure_1vv
While this reagent is only useful on aldehyde substrates, a similar bench stable reagent can be applied towards the α-oxygenation of ketones.  Treatment of cyclohexanone with N-methyl-O-benzoylhydroxylamine hydrochloride (669393) in water provides the benzoyl-protected alcohol in excellent yield, presumably through a pericyclic rearrangement of an intermediate iminium ion (Scheme 2).  A previous synthesis of this molecule from cyclohexanone by House required a laborious five-step procedure.5
scheme2v
Both acyclic and cyclic ketones undergo oxidation at mild temperatures to afford protected alcohols in good to excellent yield (Scheme 3, Figure 2).  The reagent is tolerant of existing functionality (e.g. sulfonamides, free hydroxyl groups, and unsaturation), and in the case of non-symmetrical ketones, oxidation occurs regiospecifically at the more substituted carbon.  In addition to water and DMSO, THF and CHCl3 can also be used as solvents with similar yields.
scheme3v
figure_2v
Sigma-Aldrich is delighted to add these new α-benzoylation tools to our growing arsenal of oxidation reagents.
   Product Information
Product # Product Name/Description Structure Add to Cart
669407 N-tert-Butyl-O-benzoylhydroxylamine hydrochloride, 97%
669393 N-Methyl-O-benzoylhydroxylamine hydrochloride, 97%
References:
  1. Chen, B.-C. et al. Org. React. 2003, 62, 1.
  2. (a) Brown, S. P. et al. J. Am. Chem. Soc. 2003, 125, 10808.
    (b) Zhong, G. Angew. Chem., Int. Ed. 2003, 42, 4247.
    (c) Hayashi, Y. et al. Tetrahedron Lett. 2003, 44, 8293.
  3. (a) Bøgevig, A. et al. Angew. Chem., Int. Ed. 2004, 43, 3317.
    (b) Hayashi, Y. et al. Angew. Chem., Int. Ed. 2004, 43, 1112.
    (c) Córdova, A. et al. Chem. Eur. J. 2004, 10, 3673.
  4. (a) Beshara, C. S. et al. Chem. Commun., 2005, 1478.
    (b) Beshara, C. S. et al. Org. Lett. 2005, 7, 5729.
  5. House, H. O.; Richey, F. A. J. Org. Chem. 1969, 34, 1430.