|
||||||||||||
|
||||||||||||
|
Phosgene is an extremely versatile reagent allowing easy access to isocyanates, ureas, carbamates, carbonates, acyl and alkyl chlorides.1 Many of these can be used as reactive intermediates, e.g. in peptide coupling reactions. As a dehydrating agent phosgene can also lead to isocyanides, cyanides and carbodiimides. (Scheme 1) Though highly toxic itself byproducts resulting from reactions with phosgene are harmless. When treated with alkaline solution only biocompatible salts are formed like sodium chloride or carbonate.
In cooperation with Buss ChemTech Sigma-Aldrich offers a safe and reliable phosgene generation kit giving simple access to small quantities of high purity, gaseous phosgene exactly when needed, while no transport and storage of liquid phosgene is necessary. The generator converts safe triphosgene into phosgene on demand using a catalyst.2 Phosgene generation can be stopped at any time. A total containment approach eliminates the risk that phosgene can reach the environment.
Advantages
Test the suitability of the generators in your own laboratory today with a starter kit and cartridges from Sigma-Aldrich. Figure 2 shows an application example of the starter kit. Solutions for the upscale to industrial production levels are available at BUSS ChemTech. Product Information
References:
|
||||||||||||
|
|
||||||||||||