|
||||||||
|
||||||||
IntroductionVarious optically active phosphine ligands incorporating a chiral center at phosphorus display exceptional enantiosectivities in metal-catalyzed asymmetric synthesis.1 For instance, known classes P-chiral phosphine ligands offer good to excellent enantiocontrol in Ru- and Rh-catalyzed hydrogenation reactions.2 The one limitation associated with these ligands is their sensitivity to air, which has impeded widespread applicability in bench-top chemistry. Imamoto and co-workers have addressed this deficiency through the invention of QuinoxP*, which contains an electron-withdrawing quinoxaline architecture.3 Aldrich, in our collaboration with Nippon Chemical, is pleased to offer R,R-QuinoxP* for the research market.† Advantages of the QuinoxP* Ligands:· QuinoxP* is not oxidized nor epimerized at ambient conditions in air · Enantioselectivities are outstanding for various reaction paradigms · Hydrogenations proceed under mild reaction conditions · Low catalyst loadings yield high TONs Representative Applications: Highly Asymmetric Rhodium-Catalyzed Hydrogenation
Asymmetric 1,4-additions of Arylboronic Acids Imamoto and co-workers exploited the high activity of the QuinoxP* ligand in rhodium-catalyzed enantioselective 1,4-additions of arylboronic acids to α,ß-unsaturated carbonyl substrates.3 High yields of the addition products were obtained via running the reactions between 40 and 50 ºC (Scheme 2). The exceptional enantiocontrol exerted by this Rh(I)-catalyzed system is evident when compared to the use of BINAP as the chiral ligand.6
Asymmetric Pd-catalyzed Ring Opening Imamoto and co-workers have also succeeded in developing a Pd-catalyzed C-C bond-forming reaction, which displays high enantioselectivities with both dimethyl- and diethylzinc (Scheme 3, Table 2). This alkylative ring-opening methodology entails simply premixing PdCl2(cod) and QuinoxP* for 2 hours at room temperature - leading to a highly active catalyst. This catalyst system affords excellent yields of the ring-opened products and selectivities that rival the highest reported for this transformation. These results, when combined with the outstanding methodologies presented above, indicate that QuinoxP* is useful for a broad variety of asymmetric metal-catalyzed transformations.
Product Information
References: (1) (a) Weinkauff, D. J. et al. J. Am. Chem. Soc. 1977, 99, 5946; (b) Spagnol, M. et al. Chem. Eur. J. 1997, 3, 1365; (c) Hamada, Y. et al. Tetrahedron Lett. 1997, 38, 8961; (d) Kurth, V. et al. Eur. J. Inorg. Chem. 1998, 597; (e) Mezzetti, A. et al. Organometallics 1998, 17, 668; (f) Imamoto, T. et al. J. Am. Chem. Soc. 1998, 120, 1635; (g) Mezzetti, A. et al. Organometallics 1999, 18, 1041; (h) Imamoto, T. J. Org. Chem. 1999, 64, 2988; (i) Imamoto, T. et al. Tetrahedron: Asymmetry 1999, 10, 877; (j) van Leeuwen, P. W. N. M. et al. J. Org. Chem. 1999, 64, 3996. (2) (a) Zhang, Z. et al. J. Org. Chem. 2000, 65, 6223; (b) Tang, W. et al. J. Am. Chem. Soc. 2003, 125, 9570; (c) Lei, A. et al. J. Am. Chem. Soc. 2004, 126, 1626; (d) Tang, W.; Zhang, X. Angew. Chem. Int. Ed. Engl. 2002, 41, 1612; (e) Tang, W.; Zhang, X. Org. Lett. 2002, 4, 4159; (f) Tang, W. et al. Org. Lett. 2003, 5, 205; (g) Tang, W. et al. Angew. Chem. Int. Ed. Engl. 2003, 42, 3509; (h) Xiao, D. et al. Org. Lett. 1999, 1, 1679; (i) Liu, D.; Zhang, X. Eur. J. Org. Chem. 2005, 646. †QuinoxP* ligands are sold in collaboration with Nippon Chemical for the research market (US and international patents pending) |
||||||||
|
|
||||||||