| Introduction |
| Introduction of chirality in organic molecules through the use of auxiliaries is a basic strategy in organic chemistry. Chiral benzylic alcohols are of particular interest because they often afford useful levels of stereoselection and are generally easy to cleave. The enantiomers of 1-(2,4,6-triisopropyl)phenylethanol ((R)- and (S)-Stericol®) are especially beneficial as chiral auxiliaries. |
| |
| Representative Applications |
| [2+2] Cycloadditions |
| The use of these materials in this context was first reported by B. M. de Azevedo and Greene1 in a highly stereoselective [2+2] cycloaddition of dichloroketene to a chiral enol ether prepared from this alcohol. The dichlorocyclobutanones prepared in this manner (dr ≈ 19:1) are versatile synthetic intermediates that have been used to access a variety of natural products, including lactones1, cyclopentanones2, amino acids3, and alkaloids4 (Scheme 1). |
| |
 |
| Similarly, the asymmetric [2+2] cycloaddition of dichloroketene was applied by Correia5 to chiral ene carbamate and by MacGee6 to α,β-unsaturated esters. In both cases, the Stericol® controlled reactions provided the same diastereomeric ratio (4:1), which was the highest obtained for these ene carbamate derivatives (Scheme 2). |
 |
| References: |
- de Azevedo, M. B. M.; Greene, A. E. J. Org. Chem. 1995, 60, 4940-4942.
- Kanazawa, A.; Delair, P.; Pourashraf, M.; Greene, A. E. J. Chem. Soc. Perkin Trans. 1997, 1911-1912.
- Nebois, P.; Greene, A. E. J. Org. Chem. 1996, 61, 5210-5211; Ceccon, J.; Poisson, J. F.; Greene, A. E. Synlett 2005, 1413-1416.
- Kanazawa, A.; Gillet, S.; Delair, P.; Greene, A. E. J. Org. Chem. 1998, 63, 4660-4663; Delair, P.; Brot, E.; Kanazawa, A.; Greene, A. E. J. Org. Chem. 1999, 64, 1383-1386; Pourashraf, M.; Delair, P.; Rasmussen, M. O.; Greene, A. E. J. Org. Chem. 2000, 65, 6966-6972; Rasmussen, M. O.; Delair, P.; Greene, A. E. J. Org. Chem. 2001, 66, 5438-5443; Roche, C.; Delair, P.; Greene, A. E. Org. Lett. 2003, 5, 1741-1744; Muniz, M. N.; Kanazawa, A.; Greene, A. E. Synlett 2005, 1328-1330; Roche, C.; Kadlecikova, K.; Veyron, A.; Delair, P.; Philouze, C.; Greene, A. E.; Flot, D.; Burghammer, M. J. Org. Chem. 2005, 70, 8352-8363; Ceccon, J.; Greene, A. E.; Poisson, J. F. Org. Lett. 2006, 4739-4742.
- Miranda, P. C. M. L.; Correia, C. R. D. Tetrahedron Lett. 1999, 40, 7735-7738.
- MacGee, D. I.; Mallais, T. C.; Mayo, P. D. M.; Strunz, G. M. Tetrahedron. 2006, 62, 4153-4161.
|
|
| Novel C–C bond forming reactions: Preparation of β,γ-unsaturated silyl sulfinates |
| The high level of stereoinduction achieved with Stericol® was further demonstrated by Vogel and coworkers1 in a new carbon-carbon bond forming reaction. In this process, a 1-alkoxy-1,3-diene reacted with sulfur dioxide in the presence of a Lewis acid to generate a zwitterionic intermediate, which was trapped by a trimethylsilyl enol ether to give a β,γ-unsaturated silyl sulfinate (Scheme 3). The Stericol® derived alkoxydiene gave the highest diastereomeric ratios (up to 25:1). |
 |
| References: |
- Narkevitch, V.; Schenk, K.; Vogel, P. Angew. Chem., Int. Ed. 2000, 39, 1806-1808; Bouchez, L. C.; Turks, M.; Dubbaka, S. R.; Fonquerne, F.; Craita, C.; Laclef, S.; Vogel, P. Tetrahedron 2005, 61, 11473-11487; Narkevitch, V.; Megevand, S.; Schenk, K.; Vogel, P. J. Org. Chem. 2001, 66, 5080-5093.
|
| Asymmetric oxidation of benzenesulfenates |
| In order to prepare stereodefined phophorus-sulfur chelating ligands, Drabowicz and coworkers1 studied the asymmetric oxidation of chiral benzenesulfenates to the corresponding sulfinates (Scheme 4). Among the chiral alcohols tested, the Stericol® derived benzensulfenate gave the highest diastereomeric ratio (13:1). |
 |
| References: |
- Hamel, M.; Grach, G.; Abrunhosa, I.; Gulea, M.; Masson, S.; Vazeux, M.; Drabowicz, J.; Mikolajczyk, M. Tetrahedron: Asymmetry 2005, 16, 3406-3415.
|
| Stericol® also has potential applications as an agent for resolution, deracemization, and determination of stereopurity. |
|
| Product # |
Product Name |
Structure |
Add to Cart |
| 672599 |
(R)-(+)-1-(2,4,6-Triisopropylphenyl)ethanol |
 |
|
| 672483 |
(S)-(–)-1-(2,4,6-Triisopropylphenyl)ethanol |
 |
|
| Back to Top |
|