|
QuadraPure™ and QuadraSil™ metal scavengers offer a variety of complementary solutions for efficient removal and economic recovery of precious metal contamination from products and organic or aqueous process streams. The above picture shows a typical ‘before-and-after’ treatment of a metal catalyst solution (initial metal content 1000 ppm) at room temperature, by simple agitation with a metal scavenger.
Polymer and Silica-based Metal Scavengers
QuadraPure™
The QuadraPure™ collection consists of two ranges of functionalized polystyrene beads:
- Macroporous, low-swell, robust beads for use in either in batch or flow processes.
- Microporous, solvent swellable beads with high capacity intended largely for R&D application.
The above graphic illustrates the effect of treating a metal contaminated reaction mixture with different QuadraPure™ metal scavengers. The original reaction mixture (obtained from a hydrogenation reaction using Wilkinson’s catalyst) contained ~200 ppm Rh metal. Treatment with either of the three QuadraPures™ reduced Rh contamination by >99%.
Technical Information for Download (PDF format):
1) QuadraPure™ and QuadraSil™ Presentation (2.9 MB PDF) 2) QuadraPure™ Technical Flier (197 KB PDF) 3) QuadraPure™ User Guide (381 KB PDF) 4) ChemFiles 2007 Supplement 1: Encapsulated Catalysts and Metal Scavengers (624 KB PDF)
| |
Product # |
Product Name |
Structure and Functional Group |
Examples of Metals Removed |
Add to Cart |
 |
655422 |
QuadraPure™ TU |
 |
thiourea |
Pd, Pt, Ru, Rh, Au, Ag, Cu, Hg, Pb, Cd, Ni, Co, Fe, V, Zn |
|
| 657026 |
QuadraPure™ IDA |
 |
imino diacetate |
Cu, Al, Ga, In, V, Pb, Ni, Zn, Cd, Be, Mn, Sr, Ba, Co, Fe |
|
| 657611 |
QuadraPure™ AMPA |
 |
aminomethyl phosphonic |
Fe, Cu, Ni, Al, Co, V |
|
| 668591 |
QuadraPure™ BZA |
 |
benzyl amine |
Rh, Pd, Cu, Co, Ni |
|
| 668605 |
QuadraPure™ BDZ |
 |
imidazole |
Rh, Co, Pd, Ni |
|
| 668583 |
QuadraPure™ EDA |
 |
amine |
Pd in basic media and with phosphines, Co, Ni, Rh |
|
| 668575 |
QuadraPure™ DET |
 |
thiol |
Pd in acidic media and with phosphines, Co, Fe, Ni, Rh |
|
 |
657654 |
QuadraPure™ IMDAZ |
 |
imidazolylpropyl amino |
Pd, Ru, Os, Co, Ni, V, Rh, Cu, Fe, Sn |
|
| 657662 |
QuadraPure™ MPA |
 |
mercaptophenyl amino |
Pd, Pt, Rh, Ni, Ru, Cu, Sn, Ag, Au, Cd, Hg, Pb |
|
| 657646 |
QuadraPure™ AEA |
 |
aminoethyl amino |
Pd, Rh, V, Cu, Fe |
|
| 551473 |
QuadraPure™ AK |
 |
activated ketone (2,4-butanedione) |
None. Scavenger for primary amines and hydrazines |
|
| 659428 |
QuadraPure™ Metal Scavengers Kit I (5 g samples of QuadraPure™ TU, IDA, AMPA, IMDAZ, MPA, and AEA) |
|
| 681547 |
QuadraPure™ Metal Scavengers Kit II (5 g samples of all QuadraPure™ scavengers with the exception of QuadraPure™ AK, total of 10 samples) |
|
References: 1) Ley, S. V. et al. Org. Lett. 2006, 8, 5231. 2) Jones, C. W. et al. Adv. Synth. Catal. 2006, 348, 1207. 3) Weck, M. et al. Adv. Synth. Catal. 2006, 348, 2101. 4) Pitts, M. R. et al. Org. Process Res. Dev. 2007, 11, 477. 5) Ley, S. V. et al. Org. Process Res. Dev. 2007, 11, 458.
QuadraSilTM
The QuadraSils™ scavengers offer extremely rapid removal of metal catalyst residues. The QuadraSils™ are based on high-quality spherical silica beads with well defined porosity which can be used in aqueous or organic solution and in batch or flow processes.
The image below illustrates the effect of treating a reaction mixture containing Grubbs’ catalyst in toluene (~ 595 ppm Ru). Addition of QuadraSils™ resulted in removal of >99% of the Ru metal, with QuadraSil™ AP reaching this low level in 1.5 hours.
Technical Information for Download (PDF format):
1) QuadraPure™ and QuadraSil™ Presentation (2.9 MB PDF) 2) QuadraSil™ Technical Flier (799 KB PDF) 3) QuadraSil™ User Guide (529 KB PDF) 4) ChemFiles 2007 Supplement 1: Encapsulated Catalysts and Metal Scavengers (624 KB PDF)
| Product # |
Product Name |
Structure and Functional Group |
Examples of Metals Removed |
Add to Cart |
| 679534 |
QuadraSil™ AP |
 |
amino propyl |
Pd, Ru, Rh, Cu, Fe, Co, Ni, Zn, Pb, Cd, Hg |
|
| 679526 |
QuadraSil™ MP |
 |
mercaptopropyl |
Pd (with or without phosphines), Rh, Cu, Ru, Pt, Pb, Ag, Hg |
|
| 679518 |
QuadraSil™ MTU |
 |
methylthiourea |
Pd, Rh, Cu, Ru, Pb, Fe, Co, Ag, Au, Hg, Pt |
|
| 679496 |
QuadraSil™ TA |
 |
triamine |
Pd, Rh, Co, Cu, Fe, Ru, Cd, Au, V, Zn, Pt, Pb |
|
| 680427 |
QuadraSil™ Metal Scavengers Kit I (5 g samples of each QuadraSil™) |
|
Carbon-based Scavengers for Trace Organics
QuadraPure™ C: Coming Soon!
QuadraPureTM C is a totally synthetic carbon adsorbent for removal of soluble organics from polar solvents, including water. It is composed of 0.3 to 0.8 mm mechanically-stable beads which are free-flowing and dust free. The adsorbent has a high surface area and capacity, is thermally stable, and is effective at removing a number of commonly-occurring organic contaminants including those responsible for color and odor. Potential applications include cGMP manufacture of APIs and intermediates and clean-up of macromolecules such as peptides.
| Easier to handle |
No reactor contamination or filter caking/blocking, dust free |
| Cleaner products |
Adsorption of trace contaminants, no “carry over” of particulate carbon. |
| Cleaner waste streams |
Removal of trace organic material from waste streams |
| Improved reproducibility |
Totally synthetic carbon (>99.9%) gives excellent batch-to- batch performance |
| Fast, efficient process |
Good filtration characteristics, fast flow rates |
| Process intensification |
Can be used in batch processes, loose packed beds, or as a pre-packed cartridge |
| Safer |
No dust or flammable solids generated |
Technical Information for Download (PDF format):
1) QuadraPure™ C Technical Flier (1.0 MB PDF) 2) QuadraPure™ C Application in Peptide Conjugate Purification (621 KB PDF)
| Product # |
Product Name |
Description |
Add to Cart |
| 684295 |
QuadraPure™ C |
Scavenger for trace organic impurities |