Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Chemistry > Chemical Synthesis > Technology Spotlights > Stabilized 2-Iodoxybenzoic Acid
Product Highlights
Stabilized 2-Iodoxybenzoic Acid
iodo1
Since 1994,1 2-iodoxybenzoic acid (IBX), has been well recognized as a very powerful and selective oxidizing agent. Like, Dess-Martin periodinane, IBX is an environmentally benign alternative to metal-based oxidizing agents. However, IBX is not often used, due to the fact it is and impact-sensitive explosive material, preventing its shipping and transport, as well as application in industry.2
In 2003, a stabilized formulation of IBX (SIBX) was reported3 which displayed none of the explosive properties of IBX, while maintaining excellent reactivity and selectivity. SIBX was then employed in a series of oxidations of aliphatic, allylic, and benzylic alcohols. The reactions proceeded cleanly, with yields comparable to those obtained with unstabilized IBX or Dess-Martin periodinane (Scheme 1).
scheme_1zz
Additionally, SIBX is capable of oxidative demethylation of 2-methoxyphenols in good to excellent yields. In one demonstrated case, oxidation proceeds in excellent yield without the loss of the silyl protecting group (Scheme 2).
scheme_2aa
Another report demonstrated the use of SIBX in the oxidative dearomatization of 2-alkylphenols into orthoquinols (Scheme 3).4 This route is a useful alternative to the Barton oxidation, which uses a selenium-based reagent, or the periodate-mediated Adler oxidation.
scheme_3zz
Sigma-Aldrich is pleased to introduce SIBX for your research needs.
Product Name Product # 
2-Iodoxybenzoic acid, stabilized (45 wt.% IBX)  new_icon21 661384
References:
  1. Frigerio, M.; Santagostino, M. Tetrahedron Lett. 1994, 35, 8019.
  2. Plumb, J. B.; Harper, D. J. Chem. Eng. News 1990, 68, 3.
  3. Ozanne, A. et al. Org. Lett. 2003, 5, 2903.
  4. Quideau, S. et al. ARKIVOC, 2003, 6, 106.